Thermo Fisher Scientific
Helping scientists answer questions that enable breakthrough discoveries in life sciences, materials science and industry
  • Categories
    • Accelerating Microscopy
    • Advancing Materials
    • Advancing Mining
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Examining Food
    • Identifying Threats
    • Life in the Lab
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAccelerating Microscopy / Electron Microscopy 101 / The History of the Electron Microscope
The History of the Electron Microscope
By Lauren Shaber 09.13.2018
Nobel Laureate Dr. Alan Finkel recently said, “Without microscopy, there is no modern science.” However, have you ever wondered how these instruments were created? Microscopes have come a long way since the first version was introduced centuries ago, as have the crucial discoveries scientists are now able to uncover. In particular, cryo-electron microscopy (cryo-EM) instruments are now to scientists what scalpels are to surgeons – tools of action, not just observation.
Brain cell image obtained with electron microscopy
Cryo-EM image depicts the interaction between a brain cell (blue) and a synthetic material (green), mimicking the extracellular environment.
Humans are inherently curious, and our fascination for the unknown drives us to dig into some of life’s most difficult challenges. Today’s advanced microscopes help scientists see the structure of viruses and proteins in 3D! Scientists now use these advanced microscopes and new technologies to study neurodegenerative diseases, such as Alzheimer’s, Parkinson’s, Huntington’s; as well as cancers including HIV, malaria, the Zika virus and many others. A better understanding of how proteins and viruses function can help researchers speed the path to more effective treatments and therapies.
The Beginning of Microscopy
The evolution of the microscope started almost as soon as the instrument was developed in the 1500s. As scientists in the 16th century began making strides in biology and chemistry, they knew they would need to see more than the naked eye would allow. Their solution was to use glass to bend light, thus creating the magnifying glass. But what would happen if they put multiple lenses together? In the late 1500s, two Dutch spectacle makers, Zacharias Janssen and his father Hans, took it upon themselves to find out. They stacked a number of lenses in a tube and realized that, together, they made items appear much larger than under a single magnifying glass. Because the maximum magnification was only 9x, they were more of a novelty than a scientific instrument.
Early observations performed with microscopy
Sketch of early microscopical nature discovery by Antonie Van Leeuwenhoek in 1825.
Antony Van Leeuwenhoek, a Dutch scientist and one of the pioneers of microscopy, became the first to create and use a microscope for scientific purposes. His hand-held, single-lens microscopes were made by grinding and then polishing glass balls into curved lenses, finding that the curvature allowed him to see objects up to 270x larger than with the naked eye. By comparison, microscopes using flat lenses could only see up to 50x magnification. He used his new microscope to analyze insects and eventually discover bacteria, earning him the title, “Father of the Microscope.”
To build on the power of a single-lens microscope, scientists had to find a way to reduce the focal length of the microscope while maintaining the lens diameter. If the lens was reduced too much, it would be difficult to see through and the images could become blurry. The solution: compound microscopes.
A compound microscope uses two or more lenses to enlarge an image to a higher magnification. Its basic structure consists of two parts:
1.      the objective, the lens closest to the object being viewed, and
2.      the eyepiece, the lens closest to the eye.
Combined, these new compound microscopes allow researchers see single-cell organisms, yeast and other building blocks of life in unprecedented detail, despite the images being slightly distorted because the glass was low-quality and the lenses imperfectly shaped. At their core, even today’s massive microscopes are considered compound microscopes.
From Photons to Electrons
In 1900, visible light microscopes reached their theoretic limit of resolution. Physics dictates light microscopes are limited to magnification of 500x to 1000x and a resolution of 0.2 micrometers (2,000 angstroms). Four years later, Carl Zeiss overcame this limitation and introduced the first commercial UV microscope, which had resolution twice as powerful as a visible light microscope. Yet, it wasn’t until nearly 30 years later that researchers found a way to blast through these limitations altogether.
In 1931, two German scientists, Ernst Ruska and Max Knoll, found a way to achieve a resolution greater than that of light. They stopped using light, instead realizing they could transmit electrons through a specimen to form an image. This discovery led to the first transmission electron microscope (TEM), where the electrons are pointed directly at the sample and pass through it to create the image.
Ten years later, Ruska created a similar yet different approach using a focused beam of electrons to scan a sample’s surface in a rectangular pattern to deliver information about its topography and composition. Unlike TEM, the image from this new scanning electron microscope (SEM) was created after the microscope collected and counted the scattered electrons.
TEM image of grass shrimp embryo
Transmission electron micrograph images of Palaemonetespugio (grass shrimp) embryos showing the development of an embryonic coat, from 1933.
Going Digital
In 1986, scientists in Japan introduced the digital microscope, which many claim revolutionized microscopy. They created a way to transfer the image from under the microscope to a computer for instant analysis. Nowadays, microscopes have built-in, high definition monitors, eliminating the need for an external computer to view images.
Moving from 2D to 3D with Cryo-EM
The major challenge with microscopy, even up to recent years, was recreating the fuzzy 2D images as sharp 3D structures. Over the course of nearly two decades, three researchers – Jacques Dubochet, Joachim Franck and Richard Henderson – created a technique for generating a 3D structure of the protein at an atomic level using an electron microscope. Their technique used vitrification to cool a sample to cryogenic temperatures, thereby allowing the biomolecules to retain their shape in a vacuum. This approach, called cryo-electron microscopy (cryo-EM), was awarded the Nobel Prize in Chemistry in 2017.
 Carbon nanotube observed with TEM
Image of the birth of a carbon nanotube from a cobalt ferrite nanoparticle obtained using a Thermo Scientific Krios 3Gi Cryo-TEM.
Looking back, the microscope has come a long way. From single-lens magnifying glasses to today’s massive electron microscopes, this technology is changing science. Thanks to the advancement of microscopes, we’re able to see items at the atomic level and up to 10,000,000x more than the human eye can see.
To learn more about microscopy, fill out this form to speak with an expert.
Share this article
275
SHARES
FacebookLinkedin
Storage pucks for cryo-EM grid boxes
New Pucks for Cryo-EM Grid Boxes to Simplify Sample Storage and Shipping
Cryo-EM sample preparation One of the challenges in cryo-EM ... by Edward Pryor / 04.14.2022
Read More
Cryo-EM tau protein structure studies unravel neurodegenerative disease mysteries
Advances in Cryo-EM Tau Protein Structure Studies Unravel Neurodegenerative Mysteries
From Alzheimer’s disease to dementia, neurodegenerative di... by Leah Lavery / 04.11.2022
Read More
epitope mapping of SARS-CoV-2 spike
Rapid Cryo-EM Epitope Mapping Could Enhance Antibody Drug Discovery
Historically, cryo-electron microscopy (cryo-EM) structure d... by Mazdak Radjainia / 02.24.2022
Read More
Lauren Shaber
Lauren Shaber is Sr. Communications Lead at Thermo Fisher Scientific.
Comments
  1. Regina L.Perry says
    December 5, 2020 at 8:15 am
    This was awesome information ! I will be teaching a segment of a Bible lesson about the God’s eye view of the microscopic world based on Romans chapter 1 verse 20.”For the invisible things of Him from the creation of the world are clearly seen , being understood by the things that are made, even His eternal power and Godhead, so that they are without excuse. “
    Reply
  2. Sherry Govender says
    February 10, 2021 at 1:59 pm
    I am acctually doing Grade 9 CAPS online and i needed to find out the history and discovery of the light and electronic microscope and how it is used today
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Introducing Accelerating Microscopy
University of Connecticut Unveils New Innovation Partnership Building
Privacy StatementTerms & ConditionsLocationsSitemap
© 2022 Thermo Fisher Scientific. All Rights Reserved.
Talk to us