The equation of trend line in these graphs is used to convert portable XRF readings (in unknown samples) from intensity to percent.
The coefficient of determination, the R2 value, is a measure of how closely the data sets correlate with each other, where a perfect correlation would have an R2 of 1. The correlation for Mo, Cu and Fe are 97%, 99% and 97%, respectively (see Figure 2). The m value beside x in the trendline equation indicates the accuracy of the results, with all three elements showing excellent agreement. For additional information, including the conclusions reached, please view the Using Portable XRF Instruments for Analyzing Ore Concentrates and Grade Control application note.
Leave a Reply