INFOGRAPHIC: 9 Fun Facts About Chromium
Chromium InfographicChromite, an oxide of iron, magnesium, aluminum, and chromium, is the only ore mineral of chromium. The USGS reports that “in nature, chromite deposits are generally of two major types: stratiform (layered) and podiform (pod shaped). Both types are associated with ultramafic igneous rocks. The world’s largest stratiform chromite deposits are found in South Africa, in what is known as the Bushveld complex…. Podiform deposits are found in layered igneous sequences that developed in oceanic crust below the sea floor.”  The USGS also reports on world chromium reserves and mining capacity in its Mineral Resource Program document. Read 9 Fast Facts about Chromium , Number 24 on the Periodic Table,and discover how you encounter chromium in both your kitchen and your dentist’s office.  
Chris Calam
Written by:
Chris Calam
Senior Sales Manager, Thermo Fisher Scientific
Chris Calam spent over 26 years at Thermo Fisher Scientific, focused on optical emission, X-Ray fluorescence and diffraction, X-ray for electron microscopes across the UK, in the industrial, research and academia markets.
Read more Calam, Chris