INFOGRAPHIC: 9 Fast Facts About Molybdenum
Molybdenum infographicElement number 42 on the periodic table does not occur in nature in its native state.  It is found only chemically combined with other elements, particularly sulfur, which makes it valuable as a catalyst in the refining of petroleum.  Read more Fast Facts about Molybdenum in this infographic.
Ali Somarin
Written by:
Ali Somarin, PhD, P.Geo
Chief Geologist, Thermo Fisher Scientific
Ali Somarin performed research, drafted application notes, and provided technical knowledge on the application of portable XRF in various fields (mining, oil/gas, environmental, gems) for Thermo Fisher Scientific during 2010-2015.
Read more Somarin, Ali