In previous articles, we wrote about the January 1905 discovery of the world’s largest gem-quality clear diamond, which was the “Cullinan” diamond, found in Pretoria, South Africa — a 3,106-carat diamond weighing 1.33 pounds. You can read about what happened to the Cullinan diamond below. We also wrote about other extraordinary diamond finds.
But there’s new buzz in the industry now after the Lucara Diamond Corp announced the recovery of an exceptional 2,492 carat diamond from its Karowe Diamond Mine in Botswana, and declared that this remarkable find is one of the largest rough diamonds ever unearthed. According to the announcement, this latest recovery joins an impressive roster of other significant finds from the mine, including the 1,758 carat Sewelô and the 1,109 carat Lesedi La Rona.
The stone was recovered from the processing of EM/PK(S) kimberlite, the dominant ore type targeted by the company during the first years of its underground mining operations.
Stock photo of macro image of Igneous rock specimens – natural Kimberlite mineral
Where are Natural Diamonds Usually Found?
Diamonds are most often found in kimberlite pipes, carrot-shaped, volcanic rock formations. Diamonds started out as carbon that crystallized deep in the earth under great pressure and temperature. Volcanic activity brought the diamonds to the Earth’s surface in kimberlite magma. Erosion of kimberlite deposits over many years caused the released diamonds and indicator minerals. Advancing and receding glaciers dispersed and transported the eroded materials hundreds or thousands of miles away, creating alluvial deposits in which sometimes contain diamonds. However, most diamonds are found in the kimberlite itself.
Natural diamonds, both gem and industrial quality, are the product of either open pit or underground mining, in which diamonds are contained in those kimberlite pipes, or they are extracted from alluvial deposits in riverbeds, coastal, and undersea locations. Each scenario requires an efficient and streamlined mining process to recover the most diamonds in the most economical way.
While kimberlite is the most common source of diamonds, finding it doesn’t guarantee finding diamonds. In fact, few kimberlite pipes actually prove worthwhile to pursue. Miners determine if a kimberlite pipe is worth pursuing by conducting regional sampling schemes and airborne geophysical surveys in a precise search area. Once a list of the best targets is assembled, grade analysis is done to find out if the kimberlite is sufficiently diamondiferous. Kimberlites are known to contain high concentrations of magnetite (strongly magnetic iron ore) and ilmenite (a black iron titanium oxide mineral).
Technology Used to Find Diamonds in the Mine
Portable XRF instruments provide fast acquisition of geochemical data for ore deposit modeling, easily determining elemental constituents for most natural low concentration samples, as well as high grade ore concentrates. XRF (X-ray fluorescence) is a non-destructive analytical technique used to determine the elemental composition of materials.
XRF analyzers determine the elemental composition of a sample by measuring the fluorescence (or secondary) X-ray emitted from elements in a sample when those are excited by a primary X-ray source. Each of the elements present in a sample produces a set of characteristic X-ray lines (“a fingerprint”) that is unique for that specific element, which is why XRF spectroscopy is an excellent technology for qualitative analysis and quantitative determination of material composition. (For more information about XRF, download this ebook: Portable XRF Technology for the Non-Scientist.)
Ore-grade assessment utilizing XRF technology helps manage blasting, excavation, and hauling activities, optimizing the site blend provided to the concentrator while preventing grade dilution or the erroneous transport of ore to the waste dump. One can rapidly delineate ore boundaries and quickly identify increasingly low-grade deposits as well as instantly flag grade, sub-grade, and waste.
It seems that when it comes to diamonds, XRF technology may be a miner’s best friend.
What Happened to the Cullinan Diamond?
According to History.com, the Cullinan was later cut into nine large stones and about 100 smaller ones, valued at millions of dollars. “The largest stone is called the “Star of Africa I,” or “Cullinan I,” and at 530 carats, it is the largest-cut fine-quality colorless diamond in the world. The second largest stone, the “Star of Africa II” or “Cullinan II,” is 317 carats. Both of these stones, as well as the “Cullinan III,” are on display in the Tower of London with Britain’s other crown jewels; the Cullinan I is mounted in the British Sovereign’s Royal Scepter, while the Cullinan II sits in the Imperial State Crown.”
The Royal Collection Trust notes that in its uncut state, the 10.1 x 6.35 x 5.9 cm diamond weighed 3,106 metric carats, and at this scale, coupled with its extraordinary blue-white color and exceptional clarity, made it the most celebrated diamond in the world.
Over 100 years and it still is the largest clear diamond found, but there have been plenty more of these giant sparkling colorless and colorful gems. The Cape Town Diamond Museum notes the 777 carat Millennium Star diamond as the second largest colorless diamond with the highest color rating of D in the world. The Congolese rough diamond at 890 carats was cut into a yellow-brown diamond that ended up weighing 407.48 carats. The Woyie River rough diamond weighed 770 carats and was dubbed one of the largest alluvial diamonds ever to be found. And the Sergio diamond (3167 carats), weighing slightly more than the Cullinan, is a carbonado diamond, a rare black diamond. And more recently we wrote about the Lulo Rose, a 170 carat pink diamond.
Additional Resources:
- Explore more aspects of mining in our Mining & Minerals virtual storefront
- ebook: Portable XRF Technology for the Non-Scientist
Editor’s Note: Portions of this article were excerpted from previously published articles in this blog, refreshed in February 2023, and updated again in August 2024.
The discovery of the Cullinan diamond and the subsequent cutting of the gem into multiple pieces has captured the world’s attention for over a century. However, as this post highlights, there are several other giant diamonds with unique features that have been discovered since then. What I found fascinating was the role of XRF technology in the diamond industry.
How does XRF technology help miners determine the quality of diamonds in kimberlite pipes and alluvial deposits, and what are the advantages of using this technology in the mining process?
Fourier Transform infrared (FTIR) spectroscopy can be used to analyze the impurities in diamond. Read more here:
https://www.thermofisher.cn/blog/mining/the-diamond-shortage-the-hunt-for-kimberlite-and-new-high-quality-synthetics/