Reader Asks: Do All Mountains Contain Precious Stones?
Reader Asks: Do All Mountains Contain Precious Stones?
Crystal Stone macro mineral surface, purple rough amethyst quartz crystals
Editor’s Note: A reader was recently intrigued after reading our previous article Where Did Those Gemstones Come From?, and asked: “Do all mountains contain precious stones?” We posed this question to one of our scientists and published her answer is below.
Thank you, reader, for your question.  First, I want to clarify that this article addresses precious gemstones, and not precious metals, which are also mined from the earth.
All gemstones (with the exception of non-mineral gemstones such as amber and pearl) are formed within the earth and through natural geological processes where they are raised to the surface — or close to it.  
To recap some of the information presented in the previously published blog article, these geological processes include the formation of three types of rocks: igneous, sedimentary, and metamorphic. 
  • Gemstones found in igneous rock include the quartzes (including amethyst, citrine and ametrine), the garnets, moonstone, apatite, diamond, spinel, tanzanite, tourmaline, topaz and zircon. Some of these gemstones form in pegmatites and hydrothermal veins that are genetically related to igneous rocks. According to the USGS, igneous rocks (from the Greek word for fire) form when hot, molten rock crystallizes and solidifies.
  • Sedimentary rock is the second type and gives rise to the transported minerals such as jasper, malachite, opal and zircon. Sedimentary rocks are formed when rock is worn down and the fragments are carried by water or the wind, and they are finally compressed together over time.
  • The third variety, metamorphic rock, is formed when intense underground heat or pressure changes existing rocks. The beryls (emerald, morganite and aquamarine), jade, lapis lazuli, turquoise, spinel, ruby, sapphire and zircon are formed in metamorphic rock.
(A well-written article detailing these processes can be found on the International Gem Society website.)
However, one should note that not all gemstones are considered to be “precious.” Typically, only four gemstones are called precious: diamond, emerald, sapphire and ruby. Other popular gemstones are considered to be semi-precious and include amethyst, malachite, turquoise, opal and lapis lazuli.
Some industry experts believe there is no real reason for the designation of precious vs semi-precious except for commercial purposes.  However, the USGS explains that “a natural gemstone is a mineral, stone, or organic matter that can be cut and polished or otherwise treated for use as jewelry or other ornament. A precious gemstone has beauty, durability, and rarity, whereas a semiprecious gemstone has only one or two of these qualities. A gem is a gemstone that has been cut and polished.”
The U.S. Federal Trade Commission notes that the value of gemstones  — no matter which kind — can be affected by their size, whether the stones are natural or lab-created, and whether they have undergone treatments to enhance them.  (Laboratory-created and imitation stones should be clearly identified as such.) Unfortunately, gemstone counterfeits, or synthetic gemstones, can be mistaken for the real thing. Another important factor in establishing the value of a gemstone is to know where it came from, both geologically and geographically, which can’t be determined visually.
Energy Dispersive X-Ray Fluorescence (EDXRF) is an important tool for the determination of the authenticity of colored gemstones and their geographical origin. Depending on the geological setting, precious gemstones like rubies, emeralds or sapphires from different origins often exhibit a characteristic combination of trace elements at different concentrations. As an example, identification and quantification of such elements may allow tracking an emerald down to its location of origin such as Colombia, Brazil, Afghanistan, Zambia or Zimbabwe. Similarly, the presence of certain trace elements also helps to distinguish between a valuable naturally formed gemstone (e.g. ruby) and a quasi-worthless synthetic crystal (e.g. synthetic ruby.)
Turning our attention back to natural precious stones, be aware that gemstones are not necessarily mined just from mountains.  A great example of this is the beach mining of diamonds in Oranjemund (Namibia), where loose alluvial diamonds were found along the Atlantic coastline.
So to answer the question Do All Mountains Contain Precious Stones?, it is unlikely that all mountains contain “precious” stones; but all mountains are formed by amazing geological processes that sometimes allow wonderful gemstones to be created and eventually moved to a place where they can be found and enjoyed by people around the world.
Additional Resources:
Comments
  1. Saqee Zayn says
    I found a store it’s white silver like stone it not too heavy it’s like white metal and shiny
    I was uploading a photo of it but here is no option
    Help me to identify the stone
  2. Nancy McShane says
    Probably talking about shinny silver pyrite cubes!
    If every mountain contained gemstones, there would be mines everywhere there are mountains, and the value would go down!