Thermo Fisher Scientific
Semiconductor manufacturing stories and solutions about a more connected, more autonomous, and smarter world
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceIlluminating Semiconductors / General / Looking to Increase Semiconductor Sample Success Rates?
Looking to Increase Semiconductor Sample Success Rates?
By David Akerson 12.02.2020
In our previous blog, we talked about how failure analysis has become a tedious and time-consuming job. FA engineers work with larger sample volumes and more complex device architectures, making it challenging to obtain precise data from every sample. FA engineers demand, fast, repeatable, high-quality milling and imaging to reduce bottlenecks caused by inefficient failure analysis workflows and increase time-to-data. The right instrument for the job is one that automates key workflows with high quality imaging to collect data from every sample.
Failure analysis of semiconductor samples with the Helios 5 PFIB DualBeam
Our new Thermo Fisher Scientific Helios 5 PFIB DualBeam FIB-SEM provides unique, site-specific, delayering and failure analysis workflows for advanced logic, 3D NAND, and 2.5/3D packaging. It also provides high speed, large area cross-sectioning, high-resolution TEM sample preparation, sub-nanometer low energy SEM performance, and automated deprocessing with end pointing for the shortest time to nanoscale information.
High resolution image of 3D NAND
High resolution, top down view of deprocessed 3D NAND
We also understand efficient sample analysis relies on optimum tool alignment to provide the highest performance under any circumstances. The SmartAlign feature included in the Helios 5 PFIB simplifies the process and eliminates the need to micro-monitor SEM alignments for automated delayering, TEM prep and slice and view cross-sectioning
Both automation and high-throughput, high-resolution analysis are crucial for time-strapped FA engineers and lab managers. The advanced capabilities of the Helios 5 PFIB speeds up time-to-data and increases productivity. Our webinar includes more details on how the Helios 5 PFIB enables higher sample throughput and better automation for semiconductor failure analysis, reducing your time-to-data to hours instead of days or weeks.
Subscribe to Thermo Fisher Scientific's semiconductor updates and resources
David Akerson is a Senior Global Market Development Manager for semiconductors at Thermo Fisher Scientific.
///
Advanced semiconductor packaging
Unraveling 3D Semiconductor Packaging Challenges
3D semiconductor packaging industry insights from Semicon Ko...
Read More
Nanoscale microscopy with FIB SEM
Nanoscale Microscopy with Gallium FIB SEM
Focused ion beam scanning electron microscopes for precision...
Read More
Sample preparation for removing large volumes of material fast with a FIB-SEM
Removing Large Volumes of Material Fast with FIB-SEMs
Preparing semiconductor samples for failure analysis and met...
Read More
Semiconductor ablation solution for Helios PFIB
New Ablation Solution for Advanced Semiconductor Packaging Increases Helios PFIB Throughput and Precision
Advanced semiconductor packaging calls for advanced failure ...
Read More
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Semiconductor Failure Analysis Takes Days…It Should Take Hours
3D NAND Flash Technology
Privacy StatementTerms & ConditionsLocationsSitemap
© 2023 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Go to mobile version