Capabilities
The Ion Torrent Genexus Dx Purification System is an in vitro diagnostic (IVD) instrument that automates nucleic acid extraction, purification, and quantification from patient specimens on a single platform to provide a consistent and efficient workflow solution for clinical next-generation sequencing (NGS) sample preparation. With just one touchpoint and ten minutes of hands-on time, you can go from specimen to purified and quantified nucleic acid that is ready to sequence on the Genexus Dx Integrated Sequencer.
The Genexus Dx Purification System pairs with the Genexus Dx Integrated Sequencer as part of the Genexus Dx System to deliver an end-to-end clinical NGS workflow from patient specimen all the way to the clinical report in as little as 24 hours. With just two user touch points and 20 minutes of total hands-on time for the sample preparation, library preparation, sequencing, and data analysis steps combined, this level of automation helps reduce user variability and ensure tracking of sample information to the final clinical report.
Simply load and go
Just load your sample onto the Genexus Dx Purification System and let the hands-free walkaway automation do the rest.
Built-in features to reduce errors
The on-board sample tracking and barcoding system helps to aid in faster sample processing while reducing errors.
Ordering Information
Intended Use: The Genexus Dx Purification System is a sample preparation instrument which automates nucleic acid extraction, purification, and quantification of DNA and RNA from FFPE tissue sections for use in next-generation sequencing applications. The system uses MagMAX magnetic bead-based technology to perform nucleic acid extraction. Purified nucleic acid is quantified using the onboard Qubit fluorometer which uses fluorescence to measure nucleic acid.
For In Vitro Diagnostic Use.
CN: 63521