Leverage your 4D in situ experiments using Digital Volume Correlation for 3D Visualization and Analysis Software
Linking the microstructure to the mechanical behavior is critical to the design of high-performance components for industry. 3D imaging techniques, such as lab/synchrotron X-ray tomography, optical tomography (OCT, OST), and MRI, allow you to perform in situ experiments with high spatial (down to the micron scale) and temporal resolution (within seconds). These techniques are particularly suitable for capturing complex and very short-lived phenomena, such as localization induced by heterogeneities, thermal mismatch between constituents, micro-cracking, fatigue behavior, and phase transitions. Digital volume correlation (DVC) exploits the natural texture of materials and has recently emerged as a powerful contactless, bulk strain measurement technique in experimental mechanics, materials science, and biomechanics.
Thermo Scientific Amira and Avizo Software are all-in-one image analysis platforms that allow you to compute 3D full-field displacement and strain maps from volume images acquired during the deformation process of an object. The data can be visualized and analyzed in a sophisticated way to quantify deformation-induced microstructural changes, such as strain transfer in multiphase materials, pore growth/coalescence related to fracture, and crack opening displacement (COD).
Where digital volume correlation can be applied?
Pore coalescence related to fracture of a fiber-reinforced polymer composite
The damage mechanisms of polyamide 6 (PA6) matrix reinforced with unidirectional glass fibers have been characterized using synchrotron radiation laminography on CT-like specimens.
Digital volume correlation (DVC) enables researchers to go one step further to quantify in more details the 3D interaction of a crack with the microstructure.
Data courtesy of L. Laiarinandrasana & T. Morgeneyer from Centre des Matériaux Mines ParisTech and L. Helfen from the European Synchrotron Radiation Facility; Karlsruhe Institute of Technology.
What tools Amira Software and Avizo Software provide for digital volume correlation (DVC) analysis?
  • 3D full-field continuous displacement and strain maps with high precision/accuracy from volume images acquired during a deformation process (possible precision down to 0.1 pixel and accuracy/bias down to 0.01 pixel, or less, depending on texture and noise)
  • Subset-based DVC method for large expected displacements (local method)
  • Finite element-based DVC method for continuous displacements (global method)
  • Fusion of local and global methods, allowing you to tackle a wide range of applications
  • Displacements and strains are computed in a 3D mesh, which can conform to the exact shape of the object, avoiding the need to trade off for a smaller region of interest
  • Advanced post processing:
    • Displacement and strain components visualized at grid elements (hexaedra/tetraedra) or nodes
    • Convert grid’s nodal displacement to a regular data set (3D image)
    • Displacement vectors visualized at grid elements, nodes, or on regular grid
    • Extraction of principal strains, invariants, eigenvectors, equivalent von Mises and Tresca strains
    • Iso-displacement and iso-strain mapping
    • Deformation animation from the 3D grid
    • Link experiment with simulation by creating tetrahedral mesh for FEA/CFD and applying DVC displacements at the mesh boundaries
    • Quantify deformation induced microstructural changes such as interface integrity or deformation induced porosity
    Services
    Amira-Avizo-Introductory-training_1160x600
    Introductory training
    Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
    The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
    Amira-Avizo-advanced-training_1160x600
    Advanced training
    Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
    The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
    Amira-Avizo-Custom-Dev_1160x600
    Custom development
    With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
    We can customize and expand our software solutions at various levels.
    Resources
    Showcases how Thermo Scientific Avizo Software can be used to analyze and understand materials deformation in composites. Data Courtesy: The University of Tokyo and KU Leuven
    Learn more  
    Read related scientific publication
    Leverage your 4D in-situ experiments using Avizo Software for Digital Volume Correlation.
    Pore coalescence related to fracture of a fiber-reinforced polymer composite.
    Analysis of 10ppi Aluminum open cell foam under compression. The Digital Volume Correlation analysis is based on the first three loading steps, showing the initial strain map distribution in the specimen. Data courtesy of TWI Ltd.
    Avizo Software for Digital Volume Correlation to understand the dessication process in Egyptian crocodile mummies.
    Showcases how Thermo Scientific Avizo Software can be used to analyze and understand materials deformation in composites. Data Courtesy: The University of Tokyo and KU Leuven
    Learn more  
    Read related scientific publication
    Leverage your 4D in-situ experiments using Avizo Software for Digital Volume Correlation.
    Pore coalescence related to fracture of a fiber-reinforced polymer composite.
    Analysis of 10ppi Aluminum open cell foam under compression. The Digital Volume Correlation analysis is based on the first three loading steps, showing the initial strain map distribution in the specimen. Data courtesy of TWI Ltd.
    Avizo Software for Digital Volume Correlation to understand the dessication process in Egyptian crocodile mummies.
    Products
    Avizo Software
    Materials Science
    • Support for multi-data/multi-view, multi-channel, time series, very large data
    • Advanced multi-mode 2D/3D automatic registration
    • Artifact reduction algorithms
    Thermo Scientific Amira 2D-5D visualization Software
    Amira Software
    Life Sciences
    • Explore 2D-5D bioimaging data
    • Identify and understand structures
    • Obtain statistical information
    • Share reports and stunning animations
    Style Sheet for Instrument Cards Original
    Contact us
    Style Sheet for Global Design System
    Style Sheet for Komodo Tabs