Search
Energy dispersive X-ray spectroscopy (EDS, or EDX) is a powerful analytical technique for characterizing chemical composition on the macro-, micro- and nanoscale. EDS derives compositional information from X-rays emitted when the electron beam scans over a sample, with each element identified from its characteristic X-ray line profile. EDS is used to analyze almost the entire periodic table, providing information on the distribution of the elements present. Information provided by EDS supports in-depth material characterization for a wide range of industrial and academic applications, spanning process/quality control to failure analysis and fundamental research.
Thermo Scientific ChemiSEM Technology offers a leap forward in usability, convenience, and speed of analysis. It integrates advanced elemental analysis with the electron image in real time. Within one software interface, the operator can:
ChemiSEM Technology simplifies EDS analysis by fully integrating SEM and EDS functions into a single, cohesive user interface. Based on live quantification and building on decades of expertise in EDS analysis, the technology provides elemental information quickly and easily, helping to ensure reliable results. And because ChemiSEM Technology is always on, it can dramatically shorten time to results, highlight features that would have previously gone unnoticed, and provide more complete information.
ChemiSEM Technology is designed and built to provide full control over all SEM and EDS functions within a single user interface. Experiment setup, image processing, and spectral analysis functions are intuitively grouped, making data management straightforward. And the system features a quadrant format and project tree to highlight the most important data.
For Research Use Only. Not for use in diagnostic procedures.