Arsenic • Metalloid
Primary XPS region: As3d
Overlapping regions: Ta5p1/2
Binding energies of common chemical states:
Chemical stateBinding energy As3p5/2
GaAs40.8 eV
As2O344.1 eV
As3+ (GaAs native oxide)44.8 eV
As5+ (GaAs native oxide)45.6 eV
As2O3 charge referenced to C1s peak at 284.8eV. GaAs oxides charge referenced to GaAs at 40.8eV.
Experimental information
  • As2O3 sublimes in vacuum.
  • As5+ may degrade under X-rays, forming As3+ as a by-product.
    • Minimize acquisition time where possible or collect data from several different locations on the sample (with very short acquisition times) and integrate the data.
Interpretation of XPS spectra
  • As3d peak has overlapping spin-orbit components (Δ=0.7eV, intensity ratio=0.7).
  • As LMM peaks (between 150eV-350eV) overlap with many XPS peaks from other elements.
As3d Spectrum Received GaAs
  • Shallow As implants can be profiled with 200eV or 500eV Ar+.
    • Typically, most of implant is within XPS sampling depth, but implant tail goes beyond XPS sampling depth.
Aresenic • Metalloid
Element Crystal Rhom
About this element
Symbol: As
Date of discovery: ancient times
Name origin: Greek arsenikos; Latin arsenicum
Appearance: gray
Discoverer: unknown
Obtained from: mispickel
Melting point: 1090 K
Boiling point: 886 K
Density[kg/m3]: 5727
Molar volume: 12.95 × 10-6 m3/mol
Protons/Electrons: 33
Neutrons: 42
Shell structure: 2,8,18,5
Electron configuration: [Ar]3d104s24p3
Oxidation state: ±3,5
Crystal structure: rhombohedral
Arsenic, a poisonous metalloid, is characteristically very similar to phosphorus, so much so that the element can be partially substituted for it in biochemical reactions. Arsenic and some of its compounds convert to a gaseous form and can rapidly oxidize to arsenic oxide, subliming directly upon heating. During the Bronze Age, arsenic was used as an impurity in the bronze to make the alloy harder. In Victorian times, women used arsenic as a complexion enhancer for their face. Fatalities occur when the toxic arsenic is handled improperly. Serious human and animal poisonings have occurred in recent years due to ingestion of arsenic – either directly or indirectly – from wood treated with copper arsenate. The most serious risk occurs when the treated wood is burned.
Products
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.