Search
Technology is a vital part of everyday life—inside and outside of the lab. Read stories from your colleagues, tips and tricks, and scientainment about using technology to benefit your life in the lab.
From machine learning predictions to personalized cell-based therapeutics—technologies propel cancer research.
Read about Nicole Gularte's battle against acute lymphoblastic leukemia (ALL) and the hope she found with CAR T cell therapy.
Learn about immunohistochemistry solutions for tumor detection using superior, validated antibodies.
Our latest NGS systems provide the versatility and scalability to handle multiple applications on a single platform.
What did the comedian and the cancer researcher have in common? They both needed help with their delivery. Here are three tips and tricks to help improve transfection of cancer cells.
Decreasing the serum content of the culture medium (to <10%) at the time of transfection is acceptable, but replace with complete growth medium within 4–24 hr posttransfection.
Antibiotics can be used during transfection.
Prior to flow cytometry, visualize cells under a brightfield microscope to verify dissociation following incubation with Gibco TrypLE reagent.
For Research Use Only. Not for use in diagnostic procedures.