Molecular Probes hydrazine, hydroxylamine and amine derivatives—Table 3.2
Table 3.2 Molecular Probes hydrazine, hydroxylamine and amine derivatives.
| Derivative | Hydrazines * | Hydroxylamines * | Cadaverines * | Other Amines * |
|---|---|---|---|---|
| Acridone | A6289 † | |||
| Alexa Fluor 350 | A10439 | A30627 | A30674 | |
| Alexa Fluor 405 | A30675 | |||
| Alexa Fluor 488 | A10436 | A30629 | A30676 | |
| Alexa Fluor 555 | A20501MP | A30677 | ||
| Alexa Fluor 568 | A10437 | A30680 | ||
| Alexa Fluor 594 | A10438 | A30678 | ||
| Alexa Fluor 633 | A30634 | |||
| Alexa Fluor 647 | A20502 | A30632 | A30679 | |
| Bimane | B30633 | |||
| Biotin and DSB-X biotin ‡ |
B1603, B2600, D20653 | A10550 § | A1594, B1596 | A1593, B1592, N6356 |
| t-BOC | B30300 ** | M6248 | ||
| BODIPY FL | D2371 | D2390 | ||
| BODIPY TR | D6251 | |||
| Cascade Blue | C687 | C621 | ||
| Coumarin | D355 | A191 † | ||
| Dansyl | D100 | D113 | D112 | |
| Dapoxyl | D10460 | |||
| 2,3-Diaminonaphthalene | D7918 † | |||
| Dimethoxybenzene | D1463 † | |||
| Dinitrophenyl | D1552 * | |||
| Fluorescein | C356, F121 | A10466 | A1351, A1353, A1363 | |
| Lucifer yellow | L453 | A1340 | A1339 | |
| Naphthalene | A91, A350 † | |||
| NBD | M20490 | |||
| Oregon Green 488 | O10465 | |||
| Pyrene | P101 | A6257 † | ||
| QSY 7 | Q10464 | |||
| Rhodamine | A1318 | L2424 | ||
| Texas Red | T6256 | T2425 | ||
| * Hydrazine, hydroxylamine and aromatic amine derivatives are discussed in Reagents for Modifying Aldehydes and Ketones—Section 3.3, and aliphatic amine derivatives are discussed in Derivatization Reagents for Carboxylic Acids and Carboxamides—Section 3.4, except for D1552, which appears in Probes for Lysosomes, Peroxisomes and Yeast Vacuoles—Section 12.3. † Aromatic amines used extensively for modifying aldehydes and ketones. ‡ Biotin and desthiobiotin (DSB-X biotin) derivatives are listed in Biotinylation and Haptenylation Reagents—Section 4.2. § ARP is the abbreviation for N-(aminooxyacetyl)-N'-(D-biotinoyl)hydrazine. ** B30300 requires t-BOC deprotection by trifluoroacetic acid to generate free hydroxylamine. | ||||
For Research Use Only. Not for use in diagnostic procedures.