Search
Lipofectamine® RNAiMAX is a proprietary formulation specifically developed for the transfection of siRNA and Stealth™ RNAi duplexes into eukaryotic cells. Lipofectamine® RNAiMAX provides the following advantages:
Quality Control
Lipofectamine® RNAiMAX is tested for absence of microbial contamination with blood agar plates, Sabaraud dextrose agar plates, and fluid thioglycolate medium, for absence of RNAse activity, and functionally by transfection of Stealth™ RNAi and appropriate controls into a reporter cell line.
Note: Cotransfecting DNA and RNA using Lipofectamine® RNAiMAX (see bottom of this section)
Reverse Transfection
Use this procedure to reverse transfect Stealth™ RNAi or siRNA into mammalian cells in a 24-well format (for other formats, see Scaling Up or Down Transfections). In reverse transfections, the complexes are prepared inside the wells, after which cells and medium are added. Reverse transfections are faster to perform than forward transfections, and are the method of choice for high-throughput transfection. Optimize transfections as described in Optimizing Transfections, especially if transfecting a mammalian cell line for the first time. All amounts and volumes are given on a per well basis.
Assessing Transfection Efficiency
To qualitatively assess transfection efficiency, we recommend using a KIF11 Stealth™ Select RNAi (available through www.lifetechnologies.com/rnaiexpress; for human cells, oligo HSS105842 is a good choice). Adherent cells in which KIF11/Eg5 is knocked down exhibit a “rounded-up” phenotype after 24 hours due to a mitotic arrest (Weil, D. et al., Biotechniques (2002), 33: 1244-1248); slow growing cells may take up to 72 hours to display the rounded phenotype. Alternatively, growth inhibition can be assayed after 48-72 hours.
Note: The BLOCK-iT™ Fluorescent Oligo (Cat. No. 2013) is optimized for use with Lipofectamine™ 2000, and is not recommended for Lipofectamine® RNAiMAX.
Forward Transfection
Use this procedure to forward transfect Stealth™ RNAi or siRNA into mammalian cells in a 24-well format (for other formats, see Scaling Up or Down Transfections). In forward transfections, cells are plated in the wells, and the transfection mix is generally prepared and added the next day. Optimize transfections as described in Optimizing Transfections, especially if transfecting a mammalian cell line for the first time. All amounts and volumes are given on a per well basis.
Note: For some cell lines (e.g. MCF-7 or HepG2), we recommend reverse transfections.
Note: Cotransfecting DNA and RNA using Lipofectamine® RNAiMAX
For cotransfections of plasmid DNA and Stealth™ RNAi or siRNA into mammalian cells, we recommend using Lipofectamine® 2000 (Catalog no. 11668-027), which is superior for plasmid transfections. If you want to use Lipofectamine® RNAiMAX for your cotransfections, perform a reverse transfection with the following modifications:
1a: Add 20 ng (for 24-well format) of plasmid DNA to the diluted RNAi duplex.
2: Add cells such that they will be 80-100% confluent 24 hours after plating.
To obtain the highest transfection efficiency and low non-specific effects, optimize transfection conditions by varying RNAi duplex and Lipofectamine® RNAiMAX concentrations. Test 0.6-30 pmol RNAi duplex (final concentration 1-50 nM) and 0.5-1.5 μl Lipofectamine® RNAiMAX for 24-well format. For extended time course experiments (> 72 hours), consider a cell density that is 10-20% confluent 24 hours after plating.
Note: The concentration of RNAi duplex required will vary depending on the efficacy of the duplex.
To transfect cells in different tissue culture formats, vary the amounts of Lipofectamine® RNAiMAX, RNAi duplex, cells, and medium used in proportion to the relative surface area, as shown in the table.
| Culture vessel | Relative surface area | Vol. of plating medium | Dilution medium reverse transfection | Dilution medium forward transfection | RNAi (pmol) | RNAi (nM) | Lipofect-amine® RNAiMAX2 |
|---|---|---|---|---|---|---|---|
| 96-well | 0.2 | 100 μl | 20 μl | 2 x 10 μl | 0.12-6 | 1-50 | 0.1-0.3 μl |
| 48-well | 0.4 | 200 μl | 40 μl | 2 x 20 μl | 0.24-12 | 1-50 | 0.2-0.6 μl |
24-well | 1 | 500 μl | 100 μl | 2 x 50 μl | 0.6-30 | 1-50 | 0.5-1.5 μl |
| 6-well | 5 | 2.5 ml | 500 μl | 2 x 250 μl | 3-150 | 1-50 | 2.5-7.5 μl |
| 60 mm | 10 | 5 ml | 1 ml | 2 x 500 μl | 6-300 | 1-50 | 5-15 μl |
| 100 mm | 30 | 10 ml | 2 ml | 2 x 1 ml | 12-600 | 1-50 | 15-35 μl |
1 Surface areas may vary depending on the manufacturer.
2 If the volume of Lipofectamine® RNAiMAX is too small to dispense accurately, and you cannot pool dilutions, predilute Lipofectamine® RNAiMAX 10-fold in Opti-MEM® I Reduced Serum Medium, and dispense a 10-fold higher amount (should be at least 1.0 μl per well). Discard any unused diluted Lipofectamine® RNAiMAX.