Search
| Organism Type | Pig | Epithelial-like; susceptible to vaccinia, rubella, and herpes viruses. |
| Tissues | Kidney | |
| Phenotype | Adherent | |
| Primary | no | |
| Application | Cell Culture / Growth Conditions,Protein Expression,Transient Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||