Acetyl-Histone H3 (Lys9) Antibody (710293) in FACS
Flow cytometry analysis of Histone H3 [AcK9] was done on HeLa cells. Cells were fixed with 70% ethanol for 10 minutes, permeabilized with 0.25% Triton? X-100 for 20 minutes, and blocked with 5% BSA for 1 hour at room temperature. Cells were labeled with ABfinity? Histone H3 [AcK9] Recombinant Rabbit Oligoclonal Antibody (710293, red histogram) or with rabbit isotype control (pink histogram) at 2.5 �øΩg/million cells in 2.5% BSA. After incubation at room temperature for 2-3 hours, the cells were labeled with Alexa Fluor�øΩ 488 Goat Anti-Rabbit Secondary Antibody (A11008) at a dilution of 1:400 for 30 minutes at room temperature. The representative 10,000 cells were acquired and analyzed for each sample using an Attune�øΩ Acoustic Focusing Cytometer. The purple histogram represents unstained control cells and the green histogram represents no-primary-antibody control.
Flow cytometry analysis of Histone H3 [AcK9] was done on HeLa cells. Cells were fixed with 70% ethanol for 10 minutes, permeabilized with 0.25% Triton? X-100 for 20 minutes, and blocked with 5% BSA for 1 hour at room temperature. Cells were labeled with ABfinity? Histone H3 [AcK9] Recombinant Rabbit Oligoclonal Antibody (710293, red histogram) or with rabbit isotype control (pink histogram) at 2.5 �øΩg/million cells in 2.5% BSA. After incubation at room temperature for 2-3 hours, the cells were labeled with Alexa Fluor�øΩ 488 Goat Anti-Rabbit Secondary Antibody (A11008) at a dilution of 1:400 for 30 minutes at room temperature. The representative 10,000 cells were acquired and analyzed for each sample using an Attune�øΩ Acoustic Focusing Cytometer. The purple histogram represents unstained control cells and the green histogram represents no-primary-antibody control.
Related Products
Related Images