1x1 image pixel for data collection
Gibco CTS DynaCellect Magnetic Separation System Software Download
These software downloads are for the CTS DynaCellect Magnetic Separation System (Catalog numbers: A55867 and A55868).
Software versions include all enhancements of previous releases in addition to the new features listed below. Updates will not in any way affect the performance of your CTS DynaCellect except for the specific changes outlined below. Links and release notes for all versions are included below. We recommend updating your system to the latest version for optimized performance.
Software downloads
DynaCellect GUI System Software
DynaCellect Protocol Builder Software
DynaCellect Protocol Builder cloud application
Version features and changes
New features/changes with version 1.2.0
  1. New variable feature: The new "Variable" feature allows users to customize volumes before different runs using the same isolation protocol. This allows for greater adaptability to specific needs.
  2. Removal of bead calculator: The Bead Calculator feature has been removed from the Isolation protocol to help streamline the user experience.
  3. Removal of the “Export Role” screen: The "Export Role" screen at the end of the protocol run has been removed to enhance workflow efficiency.
  4. Export run reports in clinical mode: Users can now export run reports while operating in clinical mode. This provides greater flexibility and access to important data.
  5. Enhanced bubble sensor checks: The Bubble Sensor checks have been improved to support more accurate and reliable operation.
New features/changes with version 1.1.1
  1. Fixed the alignment of the display for the input field of the network drive
  2. Fixed the incorrect inheritance of the “Last Run” timestamp when a protocol is created from a template
  3. Added the ability to export a run report as a PDF file to the network drive
  4. Fixed an issue that causes some users to encounter an insufficient disk space error when the instrument is in idle mode
  5. Added additional information in the run report (step name, elapsed step timer, and paused step timer)
  6. Fixed an issue where aborting an active run with a paused step timer does not immediately reset the step timer when the next run is started
  7. Fixed an issue with the bead calculator not returning the value based on user input
  8. Fixed a run stop timestamp inconsistency between the protocol details screen and the run report
  9. Fixed an issue where the user was unable to cancel their Open Platform Communications (OPC) certification setup or account
  10. Added the ability to sort by the last run date and time for both run histories and protocols
  11. Fixed an issue where additional spacing is observed in the expected file name after an emergency stop is performed
  12. Fixed an issue where pump speed is not reflected correctly on OPC
  13. Fixed an issue of incorrect output volume due to rounding error
New features/changes with version 1.1.0
  1. Added OPC mode in CM mode
  2. Revised workflows and improvement to OPC mode
  3. More data captured in CM mode
  4. Improved workflows in CM mode
  5. Changed run report from .txt to .pdf file type
  6. Allowed run report to be exported from run history screen
  7. Added valve status indicator during a protocol run and manual mode run
  8. Updated estimated run end time to include checking of valve before a run
  9. Allowed special characters to be used during import/export of protocols over network drive
  10. Improvement to static IP connection, run history, importing of protocols, bead removal pre-run and emergency stop
New features/changes with version 1.0.2
  1. Revised workflow in Open Platform Communications (OPC) mode
  2. Restricted guest user from accessing OPC mode
  3. Added checking of valves before a protocol run
  4. Added cleanup of all OPC profiles during instrument factory reset
  5. Added cybersecurity update
  6. Allowed magnet toggle as the sole requirement in a protocol
  7. Fixed bugs on buttons, emergency stop, prompts, and pre-run checks
Download instructions
Downloading software file
  1. Download or transfer the firmware file (shown in .update format) onto a USB memory stick
  2. Ensure the firmware file is not in any folder on the USB memory stick
Updating your CTS DynaCellect instrument
  1. Power on the instrument and log in as an admin user.
  2. Insert the USB memory stick containing the firmware update into one of the USB ports in the CTS DynaCellect instrument.
  3. In the Menu screen, touch “Settings”.
  4. In the Settings screen, touch “Maintenance & service”.
  5. In the Maintenance & service screen, touch “Software update”.
  6. If you have downloaded the firmware properly, the instrument should list all the available update packages on the USB memory stick. Please select “DynaCellect-1.0.2 update”.
  7. Touch “Select” at the bottom right of the screen to confirm that you want to upgrade.
  8. Touch “Proceed” at the bottom right of the screen to proceed to upgrade.
  9. When the upgrade process is complete, the instrument will restart automatically. When it has rebooted, you can remove your USB memory stick.
IMPORTANT: Ensure that you do not turn off the instrument when the upgrade is in progress.
1x1 image pixel for data collection