Search
IP-MS enrichment of EGFR (LFQ intensity): EGFR was enriched 23-fold from A549 lysate compared to background proteins, using the optimized IP-MS workflow with Pierce MS-Compatible Magnetic IP Kit protein A/G (Part No. 90409) and EGFR antibody (Part No. MA5-12880). The STRING database (www.string-db.org) was used to identify the protein interactor list. See more information on IP-MS verification of antibody selectivity.