Chromatography and Purification Products
Chromatography and purification products for multiple stages
Thermo Fisher Scientific offers a broad portfolio, with chromatography products that can support development from early research through large-scale manufacturing. These products are designed to align with specific workflow needs for modalities including monoclonal antibodies (mAbs), therapeutic proteins, cell and gene therapies, messenger RNA (mRNA), antibody-drug conjugates (ADCs), and vaccines.
Comprehensive product offerings to meet your workflow needs
Chromatography Resins
Chromatography resins are available for affinity, ion exchange (IEX), hydrophobic interaction chromatography (HIC), and mixed-mode applications. Formats range from early-stage evaluation tools to production-scale resins used in CGMP biomanufacturing.
CaptureSelect and POROS Technologies
CaptureSelect resins are widely used in the purification of complex biologics, including modern antibody formats, viral vectors, and therapeutic proteins. POROS resins support high-speed chromatography steps, making them suitable for use from process development through scale-up
Chromatography Columns
Pre-packed columns simplify setup by reducing the need for on-site column packing. Formats range from bench-scale to process-scale and include resins for capture, intermediate, and polishing steps in purification workflows.
96-Well Screening Plates
A 96-well plate format can help streamline resin evaluation and screening by running multiple purifications in parallel. This approach enables the comparison of binding and wash conditions early, before proceeding to larger-scale purification studies
Analytical Chromatography
Analytical chromatography columns, including affinity and non-affinity, are used in assay development and biomolecule characterization to support downstream processing decisions.
Membrane Chromatography
Membrane chromatography offers higher flow rates and shorter processing times compared to traditional packed-bed resins and can help accelerate biologics production from development to manufacturing.
DynaChrom Single-Use Chromatography System
The DynaChrom system is a single-use platform for pilot-scale chromatography. It is used in process development and pilot-scale manufacturing, aiming to create a bridge between laboratory studies and commercial purification.
Explore our full portfolio of bioprocessing products
Beyond chromatography and purification, our bioprocessing portfolio includes upstream systems, cell culture media, buffers, and single-use technologies to support end-to-end workflows.
Discover chromatography techniques
Chromatography can separate biomolecules by properties such as charge, hydrophobicity, or affinity. Depending on the molecule, different chromatography techniques may be used during capture, intermediate purification, or polishing.
Affinity chromatography
Affinity chromatography is commonly used as the capture step in downstream purification. The target biomolecule, produced in a complex mixture during upstream processing, binds specifically to a ligand on the affinity resin. The target binds tightly to the resin while most process-related impurities either flow through or are washed away. Elution is achieved by applying a buffer that disrupts the binding interaction, releasing the purified target from the column. This technique enables high selectivity and purity early in the purification process, making it one of the most efficient capture methods in bioprocessing.
Hydrophobic Interaction Chromatography (HIC)
HIC separates biomolecules based on differences in surface hydrophobicity. In biologics manufacturing, HIC is often used as a complementary polishing step alongside ion exchange chromatography (IEX), helping extend purification power by targeting properties that charge-based methods alone may not fully resolve.
Mixed-Mode Chromatography
Mixed-mode chromatography utilizes ligands that interact through multiple properties, such as charge and hydrophobicity. This enables the separation of biomolecules that are challenging to resolve with single-mode methods in fewer purification steps.
Chromatography solutions by modality
Discover purification solutions curated to align with the downstream workflows of your biotherapeutic development and manufacturing processes.
Antibody Purification
Increase yield and productivity in antibody purification by selecting the appropriate affinity and polishing chromatography resins that can achieve required selectivity with excellent recovery and broad impurity clearance efficiently and effectively.
mRNA
Chromatography in mRNA production separates full-length transcripts from impurities produced in upstream production, such as plasmid template and truncated products. High quality purified mRNA can then be packaged, such as in lipid nanoparticles, and formulated for therapeutic use.
Gene Therapy
Gene therapy often relies on viral vectors, such as adeno-associated virus (AAV) or lentivirus, to facilitate the transfer of genetic material. Chromatography separates drug product from product- and process-related impurities in both capture and polishing steps.
Plasma Fractionation
Selective chromatography technologies, such as affinity chromatography, support higher yield and purity during blood plasma fractionation, helping manufacturers extract greater therapeutic value from available plasma.
Recombinant Proteins
Recombinant therapeutic proteins—including hormones, enzymes, and fusion proteins—require chromatography-based downstream purification workflows that can vary significantly by target molecule.
Learn more about chromatography products
Chromatography resources
Chromatography Resin Selection Tool
Custom Chromatography Services
Bioprocessing Downstream Calculator
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.