Search
The Thermo Scientific Metrios 6S (S)TEM enables automated TEM metrology utilizing automated TEM grid handling and seamless data connectivity between systems within the Thermo Scientific Vulcan Automated Lab. Leveraging our expertise in these areas, the Metrios 6S (S)TEM is designed to address the distinct challenges encountered in scaling lab operations.
The Metrios 6S (S)TEM is a new-generation, fully automated instrument that enables high-volume TEM metrology. The Metrios 6S (S)TEM features newly designed hardware and machine-learning-based capabilities. As part of the Vulcan Automated Lab, SEMI standard compliant TEM grid handling and host connection are seamlessly integrated, allowing automated TEM grid transfer from the Thermo Scientific Helios 6 HXS FIB-SEM and continuous dataconnectivity with a customer-provided host network. Along with Smart Automation software, this combination provides improved TEM metrology productivity and recipe-free automation for scalable lab operations and resource optimization.
The new Metrios EFEM (Equipment Front End Module) is an innovative TEM grid handling system designed for the Metrios 6S (S)TEM as part of the Vulcan Automated Lab. It provides an automated, operator-free solution for high-throughput loading and unloading of TEM grids, seamlessly integrating with automated materials handling systems (AMHS) via overhead tracks (OHT) or autonomous mobile robots (AMR). In this workflow, samples are delivered to the Helios 6 HXS (S)TEM for creation of lamellas, which are then transferred to the Metrios 6S (S)TEM for high-precision metrology data collection.
Automated data connectivity facilitates seamless SEMI standard communication between a customer-provided Hostnetwork and the Metrios 6S (S)TEM’s workflow control software, including interfacing with the customer's materialhandling system.
The Metrios 6S (S)TEM offers sophisticated, automated metrology, delivering precise nanoscale measurements through automated image acquisition and analysis, essential for semiconductor manufacturing. Its Smart Automation software significantly reduces set-up time for new processes and sample types, ensuring ease of use and scalability by leveraging advanced machine-learning algorithms. The instrument’s flexibility allows operation in both full-auto and semi-auto modes, adapting to specific lab needs and accommodating a wide range of challenging semiconductor device types. For model training, a new web-based segmenter and object detection algorithm accelerate the time to build robust Al models. For additional flexibility, conventional recipe-based automation is also available.
TEM grid handling |
SEMI standard-compliant, operator-free grid handling via AMHS. |
Data connectivity |
SEMI standard-compliant software interface for connection to a customer-provided host network. |
Metrology |
TEM and STEM mode metrology accuracy: Less than 0.75% combined error in distortion and magnification calibration. |
Application software |
Smart Automation software allows setup of new automation routines in <4 hours for select workflows. EDS quantification using Thermo Scientific Velox Software, featuring dynamic correction of holder shadowing as a function of tilt. |
EDS |
Ultra-X detector provides high-sensitivity, windowless EDS detection with high solid angle and high cleanliness. |
Electron column |
Three-lens condenser system with indicators for convergence angle and size of the illuminated area, allowing for quantitative measurement of electron dose and illumination conditions. The S-CORR probe corrector provides sub-Ångström imaging resolution at 60 kV as specification and an order of magnitude improvement in optical stability. The S-CORR corrector simultaneously corrects A5 for all accelerating voltages. Flexible, high-tension range of 60–200 kV. Choice of ultra-stable, high-brightness Schottky field emission gun (X-FEG) or ultra-high- brightness cold field emission gun (X-CFEG) with an energy resolution of <0.4 eV. |
Stage |
Five-axis, piezo-driven Smart Stage with minimum XYZ step size of ≤20 pm. |
Holder |
High-accuracy, double-tilt holder designed for the Smart Stage. Alpha-tilt, under-goniometer software control; Alpha tilt range ±40 degrees for high- accuracy, double-tilt holder. |
Thermo Scientific Perform Services will empower your success for the lifetime of your system. Simplify your budget and maximize availability with the extraordinary value and predictable cost of ownership provided by Thermo Fisher Scientific.
Most of the Thermo Scientific Perform Service packages offer software updates and continuous access to Thermo Scientific RAPID remote service and telephone support, ensuring that our technical experts' assistance is always at your disposal. Options for consumables coverage, extended service hours, and targeted uptime can provide additional business-critical support.
At Thermo Fisher Scientific, our digital capabilities allow us to be more agile and proactive in how we support your instrument. Leveraging data and remote technologies enables us to expand our service reach, deliver an accelerated response, and optimize your system’s uptime and performance.
Our worldwide team of 1,000 service professionals is ready to assist you with technical issues or other service inquiries. Their goal is to expediently resolve any issue that might arise with your instrument.
For Research Use Only. Not for use in diagnostic procedures.