XPS instrument overview
For over 50 years, Thermo Fisher Scientific has provided surface analysis instruments to academic and industrial customers. Our instruments deliver the performance required for advanced materials science research, product development, and failure analysis. Our diverse portfolio is designed to cater to various applications, accommodating routine, high-throughput measurements, as well as complex experimental workflows.
XPS Instruments specifications
    K-Alpha System Nexsa G2 System Hypulse System
ESCALAB QXi Microprobe
XPS instrument
capabilities
Large-area XPS
Small-area XPS
XPS imaging
SnapMap  
Charge compensation system for insulator analysis
Ion source for depth profiling
fs-laser for depth profiling      
Angle-dependent XPS Optional Optional Optional
Automated sample transfer Optional
XPS instrument analytical options Raman spectroscopy   Optional   Optional
MAGCIS Ion Source   Optional Optional
Reflected electron energy loss spectroscopy (REELS)   Optional
Low energy ion scattering spectroscopy (ISS/LEIS)   Optional
UV photoelectron spectroscopy (UPS)   Optional Optional Optional
Auger electron spectroscopy       Optional
Sample preparation options   Optional Optional Optional
Inert sample transfer Optional Optional Optional Optional
Higher energy X-ray sources (HAXPES)       Optional
For Research Use Only. Not for use in diagnostic procedures.