Standard (SD) HPLC Systems
Versatility, performance and the freedom to configure
Meet the demands of your microbore, analytical, and semipreparative LC applications with a Thermo Scientific UltiMate 3000 Standard (SD) HPLC/UHPLC System. These systems feature versatility and performance with the freedom to configure the best system for your application. They are available with binary, quaternary and dual pump systems to meet different application requirements. All system components are configured to produce best quality results.
Popular standard HPLC systems-related products
Featured standard HPLC systems
Get mobile phase flexibility for your high-throughput routine or R&D separations. With a wide mixing portfolio for gradient delay volume adjustment, four-solvent gradient support, excellent flow stability and an unique choice of detection technologies, this system excels where moderately fast gradient response is acceptable to gain the advantages of improved flexibility such as in the following applications:
  • Multiple solvents
  • Extended column washes
  • Ternary or quaternary gradients
  • Method development scenarios
Increase the speed and throughput of your workflows without redeveloping existing methods. This system is ideal for routine and advanced chromatographic methods, productivity solutions, and multidimensional analyses. The dual system lets you maximize throughput, resolution, or sensitivity, and offers options for improving information yield. It is easily configured for:
  • Online-SPE
  • Orthogonal, parallel or tandem separations,
  • Inverse gradient
  • Fast application switching
  • Operation as a regular single-channel system
Discover the best HPLC systems for your applications and budget
Discover the best HPLC system for your applications and budget
Use these tools to help you decide
HPLC Systems Selection Guide
Use this tool to rank the top HPLC instruments for your applications based on your responses to simple pull-down menus.
HPLC System Total Cost of Ownership Calculator
Use this tool to estimate your cost for purchasing and running Thermo Scientific HPLC and UHPLC systems. You can compare costs between instruments.
LC Method Transfer Calculator
Use this online calculator to transfer methods from HPLC to UHPLC conditions. Calculate the new gradient table, adjust sample volumes and run times if applicable.