cartridge
Lab-quality forensic DNA profiles
Product description
Part number category
Part number
Price
RapidHIT ID ACE GlobalFiler Express 150 Sample Cartridge Kit
Consumables
400093RH
Request a Quote
RapidHIT ID ACE NGM SElect Express 150 Sample Cartridge Kit
Consumables
400095RH
Request a Quote
RapidHIT ID EXT GlobalFiler Express 150 Sample Cartridge Kit
Consumables
400127RH
Request a Quote
RapidHIT ID Primary Cartridge 250
Consumables
400118RH
Request a Quote
RapidHIT ID ACE GlobalFiler Express 125 Sample Cartridge Kit
Consumables
400121RH
Request a Quote
RapidHIT ID ACE NGM SElect Express 125 Sample Cartridge Kit
Consumables
400122
Request a Quote
RapidHIT ID EXT GlobalFiler Express 125 Sample Cartridge Kit
Consumables
400115RH
Request a Quote
RapidHIT ID EXT NGM SElect Express 125 Sample Cartridge Kit
Consumables
400117RH
Request a Quote
Description:
The IntegenX by Thermo Fisher Scientific RapidHIT ID consumables support legacy IntegenX systems.
The compact, easy-to-use IntegenX RapidHIT ID System generates lab-quality forensic DNA profiles in virtually any setting in less than 90 minutes. Processing a buccal swab is simple, requiring less than one minute of hands-on time. DNA profiling has never been this fast or this easy.
RapidHIT ID generates DNA STR profiles using NDIS-approved chemistry, developed to meet FBI Quality Assurance Standards (QAS) and criteria for Rapid DNA analysis. RapidHIT ID data formats are compatible with established DNA databases (e.g., CODIS, NDNAD, NCIDD).
  • Compact instrument footprint
  • Lab-quality DNA profiles in <90 minutes
  • Less than 1 minute of hands-on time
Specifications:
Performance
Processing time >90 minutes
Applied Biosystems GlobalFiler Express–24 markers, including expanded core CODIS loci
Applied Biosystems AmpFLSTR NGM SElect Express–17markers
Operating environment
15–35ºC (59–95ºF)
20–90% relative humidity
Altitude up to 3,000 m (9,842 ft)
100–240 VAC, 50/60 Hz, 120V, 600W