Thickness and coating weight gauges for hot and cold rolling processes
Ensure your products meet the most precise specifications
Produce quality steel products while maintaining output and efficiency with the help of our complete line of non-contact thickness gauges for hot- and cold-rolling mills that provide precise, real-time measurements during high-speed production of steel plate and sheet to help you meet the tightest tolerances while maximizing raw material use.
For zinc-coated steel sheet, our hot coating weight gauges provide fast feedback for coating control using well-established X-ray fluorescence (XRF) and our near-infrared (NIR) sensors assure uniform paint coverage.
Contact a sales representative
Featured metal thickness and coating weight gauges used in steel manufacturing
Video spotlight: Metal thickness and coating weight gauges
Metals weight and coating thickness gauge product comparison guides
Metals weight and coating thickness gauge product comparison guides
We offer a complete line of non-contact thickness gauges for hot- and cold-rolling mills that provide precise, real-time measurements that meet the tightest specifications of any application while maximizing raw material use. 
Need help choosing a metal thickness measurement or metals coating weight gauge? Use these comparison tables to see which model is right for you.
Metals Coating Weight Gauges Product Comparison Table
Metals Thickness Measurement Gauges Product Comparison Table