Search
Choose the right Thermo Scientific Raman microscope, spectrometer or analyzer for your specialized work. Because the needs of a process engineer, a manufacturing QA/QC director, or a lab researcher all differ in specific ways, we have multiple instrumentation options to meet different requirements. Ideal for use in forensics, pharmaceutical analysis, battery development, academic research and more, this selection guide can help to find the Raman analytical instrument that best fits your chemical identification and process measurement needs.
This selection guide is designed to help you decide which Raman instrument best fits your needs. Whether you need micron-level spatial imaging information about a sample, verification and validation of a bulk powder, or in-line process monitoring, we have an instrument for the task.
Microscopes |
Macro/laboratory spectrometers |
Macro/At-line Pharma and Industrial QC | Process |
Handheld | |||
![]() |
![]() |
![]() |
![]() |
![]() |
![]() |
![]() |
|
| Model | DXR3xi Imaging Microscope | DXR3 Microscope |
DXR3 SmartRaman Spectrometer |
DXR3 Flex Raman Spectrometer |
DXR3 SmartRaman+ Spectrometer | MarqMetrix All-In-One Process Raman Analyzer |
TruScan G3 Handheld Raman Analyzer |
| Design/ function | Suitable for advanced Raman imaging and high-resolution work |
Ideal for general Raman microscopy applications |
Designed for bulk sample analysis with high sensitivity and automation features |
Provides a versatile and modular Raman system for a variety of applications |
Higher-throughput at-line testing during fill/finish in Pharma manufacturing and product quality testing in industrial settings. |
Designed for process and rapid measurements, and straightforward bulk sample analysis |
Rapid material identification testing for pharmaceutical and biotechnology manufacturing quality control/quality assurance processes |
| High-level capabilities and use-case scenarios |
|
|
|
|
|
|
|
| Industries | Batteries │ Graphene │ Pharma │ Semiconductor │ Environmental │ Materials │ Polymer/Chemical │ Academic research │ Forensics |
Primary: Pharma | Biopharma Secondary: Polymer | Chemical |
Pharma/Biopharma | Oil/Gasoline | Batteries | Polymer/Chemical | Food/Beverage |
Pharma/Biopharma |
|||
| Key users | Researchers │ Manufacturing QA/QC │ Engineers |
Process engineers | Quality engineers | MSAT Scientists | Process development engineers |
Process engineers | Quality engineers | MSAT Scientists | Process development engineers |
Manufacturing QA/QC | Quality engineers | Researchers |
|||
| Lasers supported | 455 nm, 532 nm, 633 nm, 785 nm |
532 nm, 785 nm |
785 nm |
785nm |
|||
| Features & benefits |