X-ray diffraction
Meet the demand for high-precision chemical analysis
Quickly obtain detailed structural and phase information of materials using x-ray diffraction (XRD), a versatile and nondestructive analytical technique. XRD analysis provides high-performance results in a wide range of industrial and research applications including academic research, chemicals, pharmaceuticals, polymers, semiconductors, thin films, metals and minerals.
If you would like to learn more or need additional information, ask to speak to a representative now.
Request for more information ›
Featured XRD systems for material science
Get a dedicated XRD system for QA/QC, academic, and routine applications with the compact, affordable Thermo Scientific ARL EQUINOX 100 X-ray Diffractometer. This transportable bench top instrument offers true flexibility.
Experience real-time measurement in a simple yet powerful benchtop XRD analyzer. The Thermo Scientific ARL EQUINOX 1000 X-ray Diffractometer offers maintenance free, simplified operation for QA and routine XRD studies.
Experience fast, accurate measurements with the Thermo Scientific ARL EQUINOX 3000 X-ray Diffractometer. This research-grade diffraction system provides high resolution detectors, a large sample area, and choice of X-ray sources.
Perform phase identification and quantification and structural analysis of polycrystalline materials. The Thermo Scientific ARL EQUINOX 5000 X-ray Diffractometer can be customized for applications requiring a four circle goniometer.
Perform advanced analyses and combined studies in real time. The Thermo Scientific ARL EQUINOX 6000 X-ray Diffractometer provides thin layer analysis, reflectometry, micro-diffraction, residual stress analysis, and texture measurements.
Achieve reliable analysis with high reproducibility and sample throughput. Thermo Scientific ARL 9900 Pot Flux X-ray Analyzer includes XRD to analyze typical bath phases and XRF to determine total calcium content in bath samples.
Perform both chemical and phase analysis in one simple operation with the Thermo Scientific ARL 9900 Total Cement Analyzer. Achieve excellent process control and ensure consistent clinker quality. 
Achieve efficient process monitoring and control for metallurgical and mining processes with full XRF and XRD capabilities in one fast, streamlined process using the Thermo Scientific ARL 9900 X-Ray WorkStation.
Video: Benchtop X-ray Diffraction System for Fast and Dynamic Analyses
Support
Follow us