HRP (horseradish peroxidase) is an enzyme frequently used as an indicator in biochemical assays. It is used to amplify signals and enhance a target’s detectability. HRP acts on various chromogenic substrates to produce many different colors and acts on chemiluminescent substrates to produce light.
HRP is often conjugated to an antibody to detect a target molecule: the antibody specifically locates the target, and HRP, in company with a substrate, produces a detectable signal. HRP is also regularly used in double labeling techniques when researchers want to simultaneously visualize different targets in the same sample. Invitrogenanti-HRP antibodies are designed to reliably detect HRP, and each antibody is validated for use in western blot, immunocytochemistry, and ELISA.
Featured product data
Western blot analysis of HRP using an anti-HRP monoclonal antibody
Western blot analysis of HRP using an anti-HRP monoclonal antibody (Cat. No. MA5-15367) that recognizes full-length HRP recombinant protein.