Decorative image of Gibco Recombinant AOF Insulin packaging
High quality animal origin free insulin
  • A wide range of product sizes
  • Gibco quality you expect
  • No performance compromises
  • Premium value
Request a quote
Insulin remains a critical component in many cell culture media and bioprocessing applications. The drive for efficiency in these applications requires an animal origin free (AOF) insulin of the highest quality and performance while providing the flexibility and value to meet your needs from validation to scale-up.
As the exclusive supplier of Biocon insulin for cell culture applications, Thermo Fisher Scientific offers it as a raw material or blended with your custom media formulations.
A wide range of product sizesPremium value
Gibco AOF insulin is available in 1 g and 5 g sample sizes from multiple lots to simplify validation and a variety of larger pack sizes to meet larger scale demands.Gibco AOF insulin is value priced up to 50% less than major competitors.
Gibco quality assuranceNo performance compromises
Gibco insulin is the only tertiary AOF insulin available on the market and is backed by the highest Gibco quality standards and documentation customers have come to expect over the past 50 years.Our insulin meets and exceeds competitive insulin product performance in cell culture growth assays and is a critical component in many of Gibco's cell culture media products.
ProductQuantityCatalog Number
Gibco rHu AOF Insulin  sourced from Biocon1 gramA11382II
Gibco rHu AOF Insulin  sourced from Biocon5 gramA11382IJ
Request a quote
Chart illustrating viable cell density vs. days in culture
Figure 1:  Biocon rHu AOF insulin demonstrates comparable growth.  HeLa cells were passaged 3 times before starting the growth curve in CD 293 Medium.
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing.
Explore our resources
For Research Use Only. Not for use in diagnostic procedures.