mem-medium-s001563-230x195
Gibco™ Minimum Essential Media is patterned after Eagle’s Media and is well suited for the growth of a broad spectrum of mammalian cells.
Choose the Right MEM
Use the Media Formulation Tool or select below.
GlutaMAX Media… Healthier Cells Live Longer
Gibco™ GlutaMAX™ media are standard cell culture media that contain a stabilized form of L-glutamine — the dipeptide L-alanyl-L-glutamine — that prevents degradation and ammonia buildup in both adherent and suspension cultures.  Use GlutaMAX media to increase media stability, minimize toxic ammonia buildup, and maximize cell performance. Get the most from your cell cultures with GlutaMAX media.
GlutaMAX Media ›
GlutaMAX-I Supplements ›
Cell Culture with Advanced MEM Media
Growth comparisons of several cell lines grown in Advanced (Reduced Serum) Media (1–2% FBS) compared to classical media (5% FBS) show equivalent or superior cell growth, with no change in morphology.
View Our Advanced MEM Media ›
Cell Culture with Advanced MEM Media