showcase.par.10026.image.0.0.1
Technology Derived From Nature
At Life Technologies we’re committed to helping improve the human condition and the world around us, by way of breakthrough research and scientific advancements. In 2012, we launched our GeneArt® TALs custom genome editing service; a novel technology derived from Xanthomonas. The precision TAL technology has been shown to be effective in a broad range of organisms including plants, algae, and fungi. GeneArt® TALs is the first technology of its kind that’s ideal for plant research scientists, offering research use access with a clear licensing path for commercial use through the Two Blades Foundation.
Precise plant genome editing has arrived; every locus is now within your reach.
About This Plant-Based Technology
Precision TALs derived from Xanthomonas, a genus of plant pathogens. The pathogen specifically recognizes DNA of the host with a protein containing 17–18 repeats of approximately 34 amino acids each. The nucleotide specificity is conferred by amino acids 12 and 13 within each repeat. Each nucleotide in the target DNA is recognized by one repeat.
This 1-to-1 relationship can be used to specifically target any locus in the genome.
Ordering GeneArt® Precision TALs for Plants
The fastest and easiest way to design, edit, optimize, and order GeneArt® Precision TALs is through the GeneArt® portal.
Order Online
The ordering process is simple; just follow the three steps below:
  1. Select the functionality (effector domain) of your TAL from Table 1. For details, refer to the ‘TALEN functionality overview
  2. Select the product name from Table 1.
  3. Then place your order through our online design tool within the GeneArt® portal, or you can download and complete the TALs order form and email it to geneartsupport@lifetech.com.
If you have a question or need free design consultation, contact us here and we’ll be happy to assist.
We will ship you a clone with a verified, optimized sequence approximately 2 weeks after confirming your order.
Table 1: Available effector domains
Product DNA-binding domain Effector domain Functionalities Applications
Native TAL Fok1 Native Fok1 endonuclease Gene targeting
  • Silencing
  • Incorporation of exogenous DNA
Truncated TAL Fok1 Truncated Fok1 endonuclease Gene targeting
  • Silencing
  • Incorporation of exogenous DNA
Native TAL VP16 activator Native VP16 activator Activation of transcription
  • Increasing the expression level of endogenous gene isoforms
Native TAL VP64 activator Native VP64 activator Activation of transcription
  • Increasing the expression level of endogenous gene isoforms
TAL repressor Modified KRAB repressor Epigenetic repression of transcription
  • Heritable knockdown of gene expression
Native TAL MCS Native (MCS) Steric repression & custom design
  • Transient knockdown of gene expression
  • Target any locus in the genome with the effector domain of your choice
Truncated TAL MCS Truncated (MCS) Steric repression & custom design
  • Transient knockdown of gene expression
  • Target any locus in the genome with the effector domain of your choice
KRAB: Krüppel associated box; MCS: multiple cloning site; TAL: transcription activator–like (DNA-binding domain).