MagMAX Cell-free Total Nucleic Acid Kit
Isolate and purify DNA and RNA (total nucleic acid) from cell-free plasma, serum, or urine samples
MagMAX kits and reagents use magnetic bead-based technology to purify high-quality DNA and RNA from cell-free plasma, serum, or urine samples. To help save time and increase reproducibility, combine with KingFisher instruments for automated purification.
Order now
  • Purify free-circulating DNA, RNA and miRNA
  • Flexible sample input from 1.0 to 6 mL plasma
  • Elution volumes ranging from 15 to 60 μL
  • Phenol-free extraction
  • Compatible with Oncomine Assays, next-generation sequencing (NGS), qPCR, and digital PCR
Automation-ready
  • Simple 4 step protocol on KingFisher instruments
  • 90 min run on KingFisher
  • Helps reduce hands-on steps compared to leading competitive alternatives
Designed to optimize your liquid biopsy research
MagMAX kits purify high-quality nucleic acids, cell-free samples and FFPE blocks, suitable for a range of research applications, including next-generation sequencing (NGS), qPCR, and digital PCR.
  MagMAX Cell-Free Total Nucleic Acid Isolation Kit
Purify free-circulating DNA, RNA and miRNA
MagMAX Cell-Free DNA Isolation Kit
Isolate cell-free DNA in 45-60 minutes
MagMAX FFPE DNA/RNA Ultra Kit
Sequential isolation of DNA & RNA from the FFPE tissue sample
Sample volumes 1.0–6 mL plasma 100 µL–10 mL
5–40 µm standard input of slides or curls
Elution volumes 15–60 μL 15–50 µL 20–50 µL
Time per run 90 minutes total, 10–15 min hands-on time with KingFisher instruments 45–60 min 20 min for DNA, 48 min for RNA
Automation/HT
  • 50 samples for 2mL plasma input
  • 25 samples for 4mL plasma input
  • 50 samples for 2 mL plasma input
  • 25 samples for 4 mL plasma input
  • 48 samples for sequential DNA and RNA
  • 96 samples for DNA or RNA only
  Learn more Learn more
  Order now Order now Order now