Search
| Site | Enzyme |
|---|---|
| 2162 | AccI |
| 4373 | AflIII |
| 2457 | AgeI |
| 2238 | BamHI |
| 943 | BbvCI |
| 3522 | BfrBI |
| 2620 | BlpI |
| 2927 | BsiWI |
| 2778 | BsmI |
| 2856 | BspMI |
| 2018 | BsrGI |
| 3442 | DraIII |
| 2337 | EcoRI |
| 2163 | HincII |
| 2176 | HindIII |
| 184 | NdeI |
| 3524 | NsiI |
| 4373 | PciI |
| 2265 | PsiI |
| 2978 | RsrII |
| 3085 | SacII |
| 2161 | SalI |
| 4257 | SapI |
| 556 | SgrAI |
| 953 | XbaI |
| 2155 | XhoI |
| Enzyme |
|---|
| AarI |
| AleI |
| ApaI |
| AsiSI |
| AvrII |
| BbsI |
| BclI |
| BglII |
| BplI |
| BplI' |
| BsaAI |
| BsaBI |
| BstBI |
| BstXI |
| BstZ17I |
| FseI |
| FspAI |
| HpaI |
| MfeI |
| MluI |
| NcoI |
| NotI |
| NruI |
| PacI |
| PflMI |
| PmeI |
| PmlI |
| PshAI |
| PspOMI |
| PsrI |
| PsrI' |
| SanDI |
| SbfI |
| SfiI |
| SnaBI |
| SrfI |
| SwaI |
| TaqII' |
| XcmI |