Size Select II
The 15 minute size separation and recovery system for library construction
  • Size separate free adapters from ligated product 10X faster in 3 easy steps
  • Optimized for tight, consistent recovery of DNA fragments <1Kb in size
  • Purified DNA is compatible with library construction protocols of major generation 2 sequencing platforms
  • Use recovered DNA directly in cloning reactions
Order nowRequest a demo
Size select DNA in 3 easy steps
With E-Gel SizeSelect II gels, you can separate and recover DNA for library construction in 3 easy steps,
Load&#x20;select&#x20;retrieve
E-Gel SizeSelect II gels are double-comb, precast agarose gels with a twist. Load your sample into the top row and electrophorese until your band or desired size range enters the bottom row. Then, easily remove the size-selected DNA with a pipette. That’s it. No additional steps are required.
Use the Invitrogen E-Gel Power Snap System (G8300), a compact, self-contained device with a built-in power supply and blue-light transilluminator, to run and visualize E-Gel SizeSelect II agarose gels. Reverse-run functionality on the E-Gel Power Snap System (G8300) allows for capturing bands of interest even if you miss the bands when they first pass through the collection wells.
Have DNA ready for library construction 10x faster
The E-Gel SizeSelect gels allow users to separate and recover DNA for short read fragment library construction in just 15 minutes. That's 10 times faster than using conventional gels, and purification kits. E-Gel SizeSelect gels are offered in a 2% agarose concentration, and validated  for the recovery of fragments in the 150-200bp size range. Once the DNA is recovered, no additional purification step is required before proceeding to the next step in library construction. E-Gel SizeSelect system has been used successfully in these generation 2 library construction protocols:
  • ABI's SOLiD Fragment Library Construction
  • ABI's SOLiD Mate Pair Library Construction
  • Illumina's Genome Analyzer Fragment Library Construction
  • Illumina's Genome Analyzer Paired-end Library Construction
Size select DNA in 3 easy steps
With E-Gel SizeSelect II gels, you can separate and recover DNA for library construction in 3 easy steps,
Load&#x20;select&#x20;retrieve
E-Gel SizeSelect II gels are double-comb, precast agarose gels with a twist. Load your sample into the top row and electrophorese until your band or desired size range enters the bottom row. Then, easily remove the size-selected DNA with a pipette. That’s it. No additional steps are required.
Use the Invitrogen E-Gel Power Snap System (G8300), a compact, self-contained device with a built-in power supply and blue-light transilluminator, to run and visualize E-Gel SizeSelect II agarose gels. Reverse-run functionality on the E-Gel Power Snap System (G8300) allows for capturing bands of interest even if you miss the bands when they first pass through the collection wells.
Have DNA ready for library construction 10x faster
The E-Gel SizeSelect gels allow users to separate and recover DNA for short read fragment library construction in just 15 minutes. That's 10 times faster than using conventional gels, and purification kits. E-Gel SizeSelect gels are offered in a 2% agarose concentration, and validated  for the recovery of fragments in the 150-200bp size range. Once the DNA is recovered, no additional purification step is required before proceeding to the next step in library construction. E-Gel SizeSelect system has been used successfully in these generation 2 library construction protocols:
  • ABI's SOLiD Fragment Library Construction
  • ABI's SOLiD Mate Pair Library Construction
  • Illumina's Genome Analyzer Fragment Library Construction
  • Illumina's Genome Analyzer Paired-end Library Construction
Ordering information
Resources
Have questions about the E-Gel system or new to molecular biology applications? Access literature, selection guides, webinars, and additional educational resources below for more information.
Literature
Read our brochure, white papers, technical notes, and white papers to facilitate your experiments.
Videos
Watch fun and educational videos on molecular biology techniques, how-tos, and tips and tricks.
Webinars
View upcoming and recorded webinars for in-depth understanding of molecular biology techniques and resources.
Selection guide
Find our E-Gel selection table that will guide you in choosing the best products for your experiments.
Product support centers
Browse the support centers by application for technical support and troubleshooting help.
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications