Designed and engineered for performance
Regardless of which model Themo Scientific CO2 incubator you choose, you can confidently pursue your cell culture endeavors. Each incubator includes rich feature sets and options suited to your unique application needs.
Explore the details of each of our incubator families by selecting them below, or select an incubator based on its features and attributes with the CO2 Incubator selection guide.
Vios iDx CO₂ Incubators
Proven performance and premium features
Advanced incubation
Cleanroom, automated, and large capacity CO2 incubators
Heracell i CO₂ Incubators
Flexible configurations for daily cell culturing
Forma Direct Heat CO₂ Incubators
Reliable performance with contamination control
Water Jacketed CO₂ Incubators
Exceptional temperature stability
Thermo Scientific Models
Economical and easy-to-use with a range of sizes
Compare models
Model Family
Vios iDx
CO2 Incubators
Advanced Incubation
CO2 Incubators
Heracell i
CO2 Incubators
Direct Heat
CO2 Incubators
Water Jacketed
CO2 Incubators
Thermo Scientific CO2 Incubators
CO2 control
1 – 21 %, 5 – 90 % optional
Varies
0-20% at ±0.1%
0-20%
0-20%
0-20%
Relative humidity
>93 % at 37 °C
Varies
<90% @ 37 °C
Ambient to 95% @ 37 °C
<90% @ 37 °C
Varies
HEPA filtration
Checkmark outline
Varies
Checkmark outline
Checkmark outline
Checkmark outline
Varies
Sterilization cycle
Steri-Run High Heat Sterilization
Varies
180 °C High-Heat Cycle
Varies
--
Varies
Stackable
Checkmark outline
Varies
Checkmark outline
Varies
Checkmark outline
--
Copper interior option
Checkmark outline
Varies
Checkmark outline
Varies
Varies
Varies
Remote monitoring
OPC-UA
OPC-UA
Thermo Fisher Connect
Thermo Fisher Connect
Thermo Fisher Connect
--