ClipTip Pipette Tips
Worry-free pipetting is finally here with the only tip that clips
Achieve confidence with Thermo Scientific ClipTip pipette tips that lock firmly in place on Thermo Scientific F1-ClipTip manual pipettes or E1-ClipTip electronic pipettes. ClipTip pipette tips feature “clips” that lock tips in place with a light touch for a secure seal on every channel.
Tips will not loosen or fall off and will only release when ejected using minimal application pressure, making daily pipetting a totally new experience. No more banging tips on your pipette to ensure they are properly attached and sealed!
View the ClipTip Selection Tool
The ClipTip pipette tip advantage
Thermo Scientific ClipTip pipette tips provide a unique locking tip interface that allows stable, straight mounting on Thermo Scientific F1-ClipTip manual pipettes or E1-ClipTip electronic pipettes.
ClipTip security
cliptip security
ClipTip pipette tips remain sealed until you decide to eject them, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
ClipTip seal
Cliptip Seal
ClipTip interlocking technology has an interface between the tip fitting and tip that seals by clipping into place.
Consistent pipetting
consistent pipetting
Enable consistent and reproducible pipetting from user to user and channel to channel.
Key ClipTip features
  • Volumes range from 12.5μL to 1250μL
  • 96- and 384-formats available
  • Low retention available in ClipTip 20-1250 96-format tips 
  • Certified free of RNase, DNase, DNA, ATP, and endotoxins
  • Color-coded for easy identification
  • Extended length tips from 12.5µl and 300µl for better reach with specific labware formats
Select your ClipTip system
ClipTip technology is the ideal solution in which the pipette and tip form a pipetting system that increases the confidence in reproducibility, reduces forces required to attach and eject tips, and secures the best possible accuracy and precision.
Pipette
Compatible tip
Features and benefits
F1-ClipTip Manual Pipettes
  • Single and multichannel
ClipTip tips:
  • 20µl, 50µl, 200µl, 300µl, 300µl Ext, 1000µl
  • Interlocking ClipTip technology provides a secure seal on every channel
  • Light tip attachment and ejection forces
  • Locking volume adjustment prevents volume drift
  • Light-weight handle with adjustable finger rest for left- or right-handed use
E1-ClipTip Electronic Pipettes
  • Single and multichannel
  • Equalizer
96-format ClipTip tips:
  • 12.5µl* and 12.5µl Ext , 200µl, 300µl and 300µl Ext, 1000µl, 1250µl
384-format* ClipTip tips:
  • 12.5µl and 12.5µl Ext, 30µl, 125µl
  • Versatile solution for liquid handling, begin pipetting immediately with preset function or create a customized, step-based program with Matrix function
  • Interlocking ClipTip technology provides a secure seal on every channel, eliminating variability
  • Equalizer’s adjustable spacing allows rapid transfer between multiple liquid vessel formats (example: tubes to plates)
  • Electronic tip ejection and index finger operation enhances ergonomics and efficiency, reducing hand strain
  • Connect Bluetooth enabled models to My Pipette Creator application for centralized, efficient programming and data storage and sharing
*384-formats, 12.5 µl compatible with the E1-ClipTip models do not have interlock technology
ClipTip1250improved
We heard your feedback and we’ve made improvements to ClipTip 1250μl tips.
  • Thinner, pointier, stronger tip shape for more precise aspiration
  • Filter raw material changed to help improve stepping applications
  • The product catalog numbers are the same
Choose the right tip for your manual or electronic ClipTip system
Tip type
Features
Recommended use
Filtered tip
  • Formats include:
  • Low retention* for maximum sample delivery
  • Aerosol-blocking filter prevents pipette and sample contamination
  • Extended length tips available in sizes 12.5µl and 300µL
  • Ideal for use in the following applications:
    • PCR and qPCR
    • RNA and DNA purification
    • Protein biology
    • Cell culture
    • Precious samples and/or sensitive assays
  • Extended lenght tips reach the bottom of specific labware formats reducing risk of cross-contaminations
Non-filtered tip
  • Non-critical samples and assays
*All ClipTip pipette tips are low retention with the exception of 384-format and 12.5 μL models
Color 96-format volume range 384-format volume range
 Pink 10 µL / 20 µL 12.5 µL
 Purple 50 µL 30 µL
 Yellow 100 µL / 200 µL 125 µL
 Orange 300 µL  ---
 Blue 850 µL  / 1000 µL ---
 Turquoise  1250 µL  ---
Packaging type
Features and benefits
Hinged-rack
  • 96- and 384-formats
  • One-handed access to tips
  • Stable base
  • Stackable to save bench space
Reload tower
  • Ecofriendly design reduces waste by 35%
  • Saves storage and bench space
  • 96 tip format
Reload inserts
  • Blister-style reload inserts for 96-format 
  • Contains a single, sterile insert of tips that fits easily into an empty ClipTip rack
  • Empty racks sold separately
Tip type
Features
Recommended use
Filtered tip
  • Formats include:
  • Low retention* for maximum sample delivery
  • Aerosol-blocking filter prevents pipette and sample contamination
  • Extended length tips available in sizes 12.5µl and 300µL
  • Ideal for use in the following applications:
    • PCR and qPCR
    • RNA and DNA purification
    • Protein biology
    • Cell culture
    • Precious samples and/or sensitive assays
  • Extended lenght tips reach the bottom of specific labware formats reducing risk of cross-contaminations
Non-filtered tip
  • Non-critical samples and assays
*All ClipTip pipette tips are low retention with the exception of 384-format and 12.5 μL models
Color 96-format volume range 384-format volume range
 Pink 10 µL / 20 µL 12.5 µL
 Purple 50 µL 30 µL
 Yellow 100 µL / 200 µL 125 µL
 Orange 300 µL  ---
 Blue 850 µL  / 1000 µL ---
 Turquoise  1250 µL  ---
Packaging type
Features and benefits
Hinged-rack
  • 96- and 384-formats
  • One-handed access to tips
  • Stable base
  • Stackable to save bench space
Reload tower
  • Ecofriendly design reduces waste by 35%
  • Saves storage and bench space
  • 96 tip format
Reload inserts
  • Blister-style reload inserts for 96-format 
  • Contains a single, sterile insert of tips that fits easily into an empty ClipTip rack
  • Empty racks sold separately
View the ClipTip Selection Tool
Transforming daily pipetting
Watch the short video and see how innovative ClipTip technology works to transform and improve your daily pipetting.
View the ClipTip System