GIBCO® DMEM Media for Superior Quality, delivers greater reliability, consistency and improved control in mammalian cell culture.
Choose the right DMEM
Growth comparisons of several cell lines grown in Advanced (Reduced Serum) Media (1–2% FBS) compared to classical media (5% FBS) show equivalent or superior cell growth and no change in morphology.
View our Advanced D-MEM (Reduced Serum Media)
DMEM Graph
Improve cell viability and growth with GlutaMAX™ Media and Supplement
While results may vary depending on the cell line, using GlutaMAX™-I Supplement in place of L-glutamine in cell culture can improve cell viability and growth, potentially increasing productivity levels. Increase media stability, minimize toxic ammonia build-up, and maximize cell performance. Get the most from your cell cultures with GlutaMAX™ media and Supplement .