F12 Nutrient Media   GIBCO® F-12 Nutrient Mixture was formulated for single-cell plating of near-diploid Chinese Hamster Ovary (CHO) cells.
Superior quality, greater reliability and improved consistency in mammalian cell culture results in better overall efficiency and more robust data.
Choose the Right F-12 Nutrient Mixture
Improve cell viability and growth with GlutaMAXTM Media and Supplement
While results may vary depending on the cell line, using GlutaMAX™-I Supplement in place of L-glutamine in cell culture can improve cell viability and growth, potentially increasing productivity levels. Increase media stability, minimize toxic ammonia build-up, and maximize cell performance. Get the most from your cell cultures with GlutaMAX™ media and Supplement.
Mammalian Cell Culture with our line of Classical Media Products
For all your additional mammalian cell culture needs checkout our line of GIBCO® brand classical media products: