Search
Symbol: No
Date of discovery: 1957
Name origin: Alfred Nobel
Appearance: unknown
Discoverer: Nobel Institute of Physics
Obtained from: man-made
Melting point: unknown
Boiling point: unknown
Density[kg/m3]: unknown
Molar volume: unknown
Protons/Electrons: 102
Neutrons: 157
Shell structure: 2,8,18,32,32,8,2
Electron configuration: P[Rn]5f147s2
Oxidation state: unknown
Crystal structure: unknown
Named after Alfred Nobel, nobelium was first synthesized by a group of scientists (Albert Ghiorso, Glenn T. Seaborg, Torbørn Sikkeland, and John R. Walton) working at the Lawrence Radiation Laboratory in Berkeley, California in 1958. With a half-life of about 58 minutes, nobelium’s most stable isotope is nobelium-259. The primary decay mode prior to the nobelium-259 isotope is alpha emission, and the primary mode after it is spontaneous fission. Nobelium has no known uses outside the laboratory because little is known about the element and only small quantities of it have ever been produced.
Products
最適なシステム性能をお届けするため、当社は国際的なネットワークで、分野ごとのサービスエキスパート、テクニカルサポート、正規交換部品などを提供しています。