Barium • Alkaline Earth Metal
Primary XPS region: Ba3d
Overlapping regions: Co2p
Binding energies of common chemical states:
Chemical stateBinding energy Ba3d5/2
BaCO3/BaO780.0 eV
BaSO4780.1 eV
Experimental information
N/A
Interpretation of XPS spectra
  • Ba3d region has well-separated spin-orbit components (Δ=15.3eV).
    • Loss features may be observed in the Ba3d reigion at 791eV and 806eV.
  • Direct overlap between Ba3d and Co2p peaks.
    • Although Ba3d / Co2p peaks have strong overlap, Ba3d spin-orbit components have a 3:2 intensity ratio compared to a 2:1 ratio for Co2p.
    • Check for the weaker Ba4d peak, if barium concentration is high enough.
Element Crystal bcc
About this element
Symbol: Ba
Date of discovery: 1808
Name origin: Greek barys
Appearance: silver
Discoverer: Sir Humphrey Davy
Obtained from: barytine, whiterite
Melting point: 998 K
Boiling point: 1413 K
Density[kg/m3]: 3510
Molar volume: 38.16 × 10-6 m3/mol
Protons/Electrons: 56
Neutrons: 81
Shell structure: 2,8,18,18,8,2
Electron configuration: [Xe]6s2
Oxidation state: 2
Crystal structure: cubic body centered
Primary uses for barium are in fireworks, vacuum tubes, fluorescent lamps, and sparkplugs. Barium, meaning “heavy” from the Greek word barys, was first discovered by C. Scheele in 1774. It is extremely difficult to obtain this metal in its pure form because of the element’s nature to quickly oxidize in air. This metal is also extremely reactive with water or alcohol. Being that water or acid-soluble barium compounds are extremely poisonous, the compounds should be handled with great care. Because barium sulfate does not dissolve and will completely rid from the digestive tract, it can be used in medicine. This compound is often used in radiography studies of human swallowing. Small amounts of barium compounds are used in paints and glassmaking.
Barium • Alkaline Earth Metal
Products
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.