Search
Join the conversation
Primary XPS region: C1s
Overlapping regions: Ru3d, Sr3p1/2, K2p
Contents of carbon section:
Binding energies of common chemical states:
| Chemical state | Binding energy C1s |
|---|---|
| C-C | ~284.8 eV |
| C-O-C | ~286 eV |
| O-C=O | ~288.5 eV |
Charge referenced to adventitious C1s, C-C peak at 284.8eV.
Adventitious carbon contamination is commonly used as a charge reference for XPS spectra.
A few nanometers of adventitious carbon on a sample can dramatically affect the survey spectrum of that sample.
Binding energies of common chemical states:
| Chemical state | Binding energy C1s |
|---|---|
| C-C | 284.8 eV |
| C=C | ~284.5 eV |
| C-O | ~286 eV |
| C=O | 288-290 eV |
| C*-CF | ~285.5 eV |
| CF2 | ~292 eV |
| CF3 | ~294 eV |
| C-N | ~286 eV |
Binding energies of common chemical states:
| Chemical state | Binding energy C1s |
|---|---|
| sp2 carbon | ~284.1 eV |
| sp3 carbon | 284.8 eV |
Functionalized graphene will have a complex C1s spectrum, containing sp2 and sp3 components.
It is generally not possible to uniquely peak fit mixed sp2/sp3 spectra.
None
Binding energies of common chemical states:
| Chemical state | Binding energy C1s |
|---|---|
| Metal carbide | ~283 eV |
| Metal carbonate | 288–290 eV |
Symbol: C
Date of discovery: Ancient times
Name origin: Latin carbo
Appearance: may be black
Discoverer: unknown
Obtained from: burning with insufficient oxygen
Melting point: 3773.15 K
Boiling point: 5100.15 K
Density[kg/m3]: 2267
Molar volume: 5.29 × 10-6 m3/mol
Protons/Electrons: 6
Neutrons: 6
Shell structure: 2,4
Electron configuration: [He]2s22p2
Oxidation state: 4,2
Crystal structure: hexagonal
Carbon has been known since ancient times when it was produced by burning organic material in the presence of insufficient oxygen. Four allotropes of carbon are known: amorphous (e.g., charcoal and soot), graphite, diamond, and fullerenes. Carbon’s small size allows it to form multiple bonds with many other small atoms, including carbon atoms, and is prevalent in a large number of chemical compounds. Carbon-based compounds are the basis for all living systems and comprise the structure of fossil fuels in the form of hydrocarbons. Several carbon isotopes are important. The International Union of Pure and Applied Chemistry designated the isotope C-12 as the basis for atomic weights, while the unstable isotope C-14, with a half-life of 5700 years, is used for carbon dating.
Products
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.