Search
Join the conversation
Primary XPS region: N/A
Overlapping regions: N/A
Binding energies of common chemical states: N/A
N/A
Symbol: H
Date of discovery: 1766
Name origin: Greek hydro and gennan
Appearance: colorless
Discoverer: Henry Cavendish
Obtained from: mines, oil, gas wells
Melting point: 14.01 K
Boiling point: 20.28 K
Density[kg/m3]: 0.08988
Molar volume: 11.42 × 10-6 m3/mol
Protons/electrons: 1
Neutrons: 0
Shell structure: 1
Electron configuration: 1s1
Oxidation state: 1,-1
Crystal structure: hexagonal
Hydrogen is the most abundant element in the universe, making up 75% of matter by mass and over 90% by number of atoms. This element is found in great quantities in stars and gas giant planets. Lighter than air, hydrogen had been used as a lifting agent in balloons and airships. However, due to its risk of fire, its use has been discontinued following the Hindenburg disaster in 1937. Hydrogen isotopes are used in the production of hydrogen bombs. In the solar wind, hydrogen’s plasma interacts with the earth’s magnetosphere, giving rise to Birkeland currents and the aurora borealis.
Products
最適なシステム性能をお届けするため、当社は国際的なネットワークで、分野ごとのサービスエキスパート、テクニカルサポート、正規交換部品などを提供しています。