Indium • Other Metal
Primary XPS region: In3d
Overlapping regions: N/A
Binding energies of common chemical states:
Chemical stateBinding energy In3d5/2
In metal443.8 eV
In2O3444.0 eV
Experimental information
  • Smaller binding energy shifts of some indium compounds compared to indium metal.
    • Chemical state differentiation can be difficult with XPS only.
    • Collect principal In MNN peak as well as In3d.
    • Bigger chemical shifts observed for In MNN compared to In3d.
  • See trace In if used as powder mounting foil.
Interpretation of XPS spectra
  • In3d region has well-separated spin-orbit components (Δmetal=7.6eV).
    • Peaks have asymmetric peak shape for metal.
    • Loss features are observed to higher binding energy side of 3d3/2 spin-orbit component for In metal.
  • In3d peaks may also show asymmetry for indium oxide, if other compounds are present (e.g. hydroxide) or there are vacancies/defects in the oxide lattice.
  • Small or negligible binding energy shifts for compounds, such as oxides.
    • In3d peaks broaden with respect to metal peaks for compounds.
    • Subtle shifts in CIGS samples.
    • Use X-ray induced In MNN Auger peaks to aid chemical state assignment.
Element Indium XPS spectra
element_indium
About this element
Element Crystal tet
Symbol: In
Date of discovery: 1863
Name origin: Indigo spectrum
Appearance: silverish
Discoverer: Ferdinand Reich
Obtained from: zinc refining
Melting point: 430 K
Boiling point: 2345 K
Density[kg/m3]: 7310
Molar volume: 15.76 × 10-6 m3/mol
Protons/Electrons: 49
Neutrons: 66
Shell structure: 2,8,18,18,3
Electron configuration: [Kr]4d105s25p1
Oxidation state: 3
Crystal structure: tetragonal
Most elements were discovered while scientists searched for other materials, and indium is no exception. This very soft, silvery-white metal has a bright luster and emits a high-pitched “cry” when bent. One of the first major applications for indium was as a coating for bearings on high-performance aircraft during World War II. Later, tin-doped indium oxide, transparent and colorless in thin films, became a main component in liquid crystal, flat panel and plasma displays. Not surprisingly the demand for indium has risen dramatically, and lower-cost alternatives, such as carbon nanotubes and conducting polymers, are being studied.
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.