Cobalt • Transition Metal
Primary XPS region: Co2p
Overlapping regions: Co LMM, Ba3d
Binding energies of common chemical states:
Chemical stateBinding energy Co2p3/2
Co metal778.2 eV
Co3O4779.7 eV
CoO779.7 eV
Experimental information
  • Strong overlap between Ba3d and Co2p XPS peaks.
    • Check for Co LMM Auger peak (713eV) or Ba4d peak (85eV) for confirmation.
  • Cobalt oxides are readily reduced by Ar+ sputtering.
    • Use lowest possible ion beam energy when attempting to clean oxide sample surface or during depth profiling.
      • Minimizes (but does not completely prevent) reduction effect.
Interpretation of XPS spectra
  • Co2p peak has significantly split spin-orbit components (Δmetal=14.99eV)
  • Co2p region overlaps some of its own weaker Co LMM Auger peaks and care must be taken not to misinterpret the peak as an XPS peak.
  • Co2p peaks have asymmetric shape for metal.
    • A well-resolved Co2p spectrum for metal shows complex structure.
High Resolution Co2p3/2 Spectrum Co Metal
  • Possible to distinguish Co oxidation states using satellite features of Co2p spectrum.
    • Co(II) has observable satellite features ~786eV.
    • In the case of Co3O4, which is actually a mixed oxidation state of Co(II) and Co(III), expect to see some satellite features due to the 2+ and 3+ states.
Co2p Spectra Co Oxides
General comments
  • Carbon may be implanted into cobalt metal forming carbides, e.g., during argon ion depth profiling.
Get materials science and electron microscopy updates
Cobalt • Transition Metal
Element Crystal Hex
About this element
Symbol: Co
Date of discovery: 1737
Name origin: German kobalt or kobold
Appearance: silver
Discoverer: George Brandt
Obtained from: arsenic, oxygen, sulfur, cobatine
Melting point: 1768 K
Boiling point: 3143 K
Density[kg/m3]: 8900
Molar volume: 6.67 × 10-6 m3/mol
Protons/Electrons: 27
Neutrons: 32
Shell structure: 2,8,15,2
Electron configuration: [Ar]3d74s2
Oxidation state: 2,3
Crystal structure: hexagonal
Frequently, cobalt is associated with nickel because both elements have characteristic ingredients of meteoric iron. This solid ferromagnetic silver-white element was known in ancient times for its compounds, but its discovery was credited to G. Brandt between 1730 and 1737 when he was able to show that cobalt colors glass a rich blue. In small amounts, cobalt is an essential element for humans and many other living organisms, and it is also a central component of vitamin B-12 or cobalamin. Cobalt should be handled with care because of its toxicity and its risk factor in nuclear confrontation. Exposure to cobalt-60, a powerful gamma ray emitter, may cause cancer.
Products
Nexsa G2 XPS
  • 미세 초점 X-선 소스
  • 독자적인 다중기법 옵션
  • 단일 원자 & 클러스터 이온 깊이 프로파일링을 위한 듀얼 모드 이온 소스
기기 카드 원본 스타일시트
K-Alpha XPS
  • 고분해능 XPS
  • 빠르고 효율적이며 자동화된 워크플로우
  • 깊이 프로파일링을 위한 이온 소스
ESCALAB Xi+ XPS
  • 높은 스펙트럼 분해능
  • 다중 기법 표면 분석
  • 광범위한 시료 준비 및 확장 옵션
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
재품 표 사항에 대한 스타일시트
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
재품 표 사항에 대한 스타일시트
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
재료 과학을 위한 전자 현미경
서비스
최적의 시스템 성능을 보장하기 위해 세계 수준의 현장 서비스 전문가 네트워크, 기술 지원 및 인증된 예비 부품을 제공해드립니다.