Search
Primary XPS region: Sc2p
Overlapping regions: Ta4p3/2, Cd3d5/2, N1s, Ge LMM, Se LMM
Binding energies of common chemical states:
| Chemical state | Binding energy Sc2p3/2 |
|---|---|
| Sc metal | 398.5 eV |
| Sc2O3 | 401.8 eV |
| ScO(OH) | 402.7 eV |
ScO(OH) referenced to Sc2p metal peak. Sc2O3 referenced to adventitious C1s peak at 284.8eV.
Experimental information
N/A
Interpretation of XPS spectra
References
About this element
Symbol: Sc
Date of discovery: 1878
Name origin: Latin Scandia
Appearance: silver
Discoverer: Lars Nilson
Obtained from: thortveitite, wiikite
Melting point: 1814 K
Boiling point: 3103 K
Density[kg/m3]: 2985
Molar volume: 15.00 × 10-6 m3/mol
Protons/Electrons: 21
Neutrons: 24
Shell structure: 2,8,9,2
Electron configuration: [Ar]3d14s2
Oxidation state: 3
Crystal structure: hexagonal
A rare transition element, scandium was discovered in 1878 by L. Nilson in Scandinavia. It occurs in only trace amounts on earth, though it is much more abundant in the sun and other stars. Although scandium can develop a slightly yellow or pink tinge upon exposure to air, it exists in its standard state as a soft, silvery white metal. Scandium’s high melting point and lightweight characteristics make it of interest to the aerospace industry and designers of sports equipment. Current applications of scandium include use in bicycle frames and in the production of high intensity light when combined with oxygen.
Products
Para garantir o desempenho ideal do sistema, fornecemos acesso a uma rede de especialistas em serviços de campo, suporte técnico e peças de reposição certificadas.