RediPlate Assay Kits—Table 10.3
Table 10.3 RediPlate Assay Kits.
RediPlate Assay Kit | Cat. No. | Assay Details |
---|---|---|
EnzChek Caspase-3 Assay Kit (Detecting Peptidases and Proteases—Section 10.4) | R35100 | A direct fluorescence-based assay for detecting caspase-3 and other DEVD-selective protease activities (e.g., caspase-7) in cell extracts and purified enzyme preparations. Caspase activity is indicated by increasing green fluorescence (excitation/emission maxima ~496/520 nm). |
EnzChek Protease Assay Kit *red fluorescence* (Detecting Peptidases and Proteases—Section 10.4) | R22132 | A direct fluorescence-based assay for detecting metallo-, serine, acid and sulfhydryl proteases and their inhibitors. Protease activity is indicated by increasing red fluorescence (excitation/emission maxima ~589/617 nm). |
EnzChek Tyrosine Phosphatase Assay Kit (Detecting Enzymes That Metabolize Phosphates and Polyphosphates—Section 10.3) | R22067 | An assay for monitoring protein tyrosine phosphatases (PTP) and for screening PTP inhibitors and modulators. PTP activity is indicated by increasing blue fluorescence (excitation/emission maxima ~358/452 nm). |
EnzChek Serine/Threonine Phosphatase Assay Kit (Detecting Enzymes That Metabolize Phosphates and Polyphosphates—Section 10.3) | R33700 | An assay for monitoring serine/threonine (Ser/Thr) phosphatases and for screening Ser/Thr inhibitors and modulators. Ser/Thr phosphatase activity is indicated by increasing blue fluorescence (excitation/emission maxima ~358/452 nm). |
RiboGreen RNA Quantitation Kit (Nucleic Acid Quantitation in Solution—Section 8.3) | R32700 | An assay for accurately quantitating low levels of RNA (1 ng/mL) with a broad dynamic range—over three orders of magnitude using two different dye concentrations. When bound to RNA, the RiboGreen reagent exhibits excitation/emission maxima of ~500/520 nm. |
For Research Use Only. Not for use in diagnostic procedures.