Determine the IC50 of a control compound
Materials required
Product or reagent Cat. No. (size) for Src assay
1 μg/mL Kinase
Prepared in Step 1.2  (Detailed assay procedure)
5X Kinase Buffer APrepared in Step 1.1  (Detailed assay procedure)
Kinase Tracer 236PV5592 (25 μL in 100% DMSO)
Eu anti-His AntibodyPV5596 (25 μg)
Master staurosporine dilution seriesPrepared prior to Step 1.1 (Compound serial dilutions)
Important:   Prior to use, antibodies used in LanthaScreen® assays should be centrifuged at approximately 10,000 x g for 10 minutes. Pipet the solution needed for the assay carefully by aspirating from the top of the solution.
Note: The detailed protocol for Src replaces Eu-anti-His with Eu-streptavidin and biotin-anti-His. For simplicity, this protocol uses Eu-anti-His only.
Note:   Antibody centrifugation is required to remove aggregates whose Tb or Eu donor signals can disrupt data analysis.
Quick reference protocol
Binding Assay—15 μL final
Step 1
Add 5 μL intermediate dilution of 3X test compound.
Step 2Add 5 μL of 3X kinase/3X antibody mixture.
Step 3Add 5 μL of 3X tracer.
Step 4Mix, cover, and read plate after 1 hour.
Final Assay conditions
Binding Assay
(15 μL final)             
Test compound
Dilution series
Kinase5 nM (typical, check protocol)
Anti-tag Antibody2 nM
Tracer~Kd*
Buffer A
1X
*~Kd for Src is 100 nM.
TOP
Detailed Assay Procedure
(3.1)   3X compound: Prepare the 100X serial dilutions of compound as described in the “Things to know before starting” section. From these master dilutions of inhibitor in 100% DMSO, prepare 3X intermediate dilutions. An example using two columns of a 96-well plate is shown. The 96-well plate is used only as a convenient vessel for preparing the intermediate dilutions (Figure 12).
Intermediate dilution guide 
Step 1Add 162 μL 1X Buffer A to 2 columns of a 96-well plate
Step 2Transfer 5 μL control inhibitor or test compound from 100X master dilution series stock, transfer from strip A to column 1 and strip B to column 2.
Step 3Mix, either with a plate shaker or 20 μL multichannel pipette.
(3.2) Transfer 3X compounds to assay plate: An example using three replicates is shown. An 8-channel pipette is used to transfer 5 μL of the intermediate 3X dilution in the 96-well plate to the 384-well plates as shown in Figure 12. Columns 1, 2, and 3 of the 384-well plate are the 3 replicates. Column 1 of the intermediate stock in the 96-well plate is transferred to alternate rows of the 384-well plate: rows A, C, E, etc. Column 2 of the intermediate stock is transferred to rows B, D, F, etc of the 384-well plate. A 16-point dilution series is created.
(3.3) Positive controls: Add 5 μL of the highest 3X compound concentration to each well in the top half of column 4 (rows A–H, see Figure 13).
(3.4) Negative controls: Prepare a 3% solution of DMSO by adding 6 μL of DMSO to 194 μL of 1X Kinase Buffer A. Add 5 μL to each well in the bottom half of column 4 (rows I–P, see Figure 13). The final concentration of DMSO in all of the wells should be 1%.
(3.5) 3X Tracer: Prepare 1 mL tracer solution in 1X Kinase Buffer A at 3X the desired final assay concentration. In the Src example, the detailed protocol states that the Kd for Tracer 236 was determined to be 178 nM, and a tracer concentration of 100 nM was chosen for the experiment. Under these conditions, a high Z’-factor of 0.72 was obtained while keeping the tracer concentration close to the Kd value. Tracer 236 is supplied at 50 μM.
To prepare 3X Tracer 236:
Stock: 50 μM = 50,000 nM
1X tracer = 100 nM (from detailed protocol)
3X tracer = 300 nM
Sample calculation for 1,000 μL:
   1X tracer =100nM   3X tracer = 300 nM 
 V1  x  
C1=V2   x  
    C2
   [Initial]   [Final 3X]
 Tracer 236V1  x  50,000
nM
=1000 μL  x  300nM
V1 = 6 μL      
Buffer:
Your calculations:
 V1  x  
C1=V2   x  
    C2
   [Initial]   [Final 3X]
Tracer 236V1  x  ____nM=1000 μL  x  ____nM
V1 = _____ μL      
Note: protocols
(3.6) 3X Kinase/3X Antibody: Prepare 3X kinase/3X antibody solution at 15 nM kinase and 6 nM antibody. In the Src example, the kinase is supplied at 0.58 mg/mL and the Eu-anti-His antibody is supplied at 0.25 mg/mL, or 1,700 nM. The kinase concentration needs to be converted from mg/mL to nM. The molecular weight for each kinase can be found on its COA.
Src has a molecular weight of 62.3 kDa; therefore, 62.3 μg = 1 nmol.
0.58 mg   x   1,000 μg    x     1 nmol      x   1,000 mL    =    9,300 nmol       =     9,300 nM
 V1xC1=V2xC2
   [Initial]   [Final 3X]
Kinase
V1x9,300 nM=1,500 μLx15 nM
V1 = 2.4 μL      
AntibodyV1x1,700 nM=1,500 μLx6 nM
 V1 = 5.3 μL      
Your calculations:
  mg1,000 μg nmol
nmol1,000 mLnmol __nM
 V1xC1=V2xC2
   [Initial]   [Final 3X]
Kinase
V1x___ nM=1,500 μLx15 nM
V1 = __  μL      
AntibodyV1x___ nM=1,500 μLx6 nM
 V1 = __ μL      
Buffer:
(3.7) Assay plate setup: Add 5 μL of 3X kinase/3X antibody solution and 5 μL of 3X tracer solution to each well of the assay plate (plate already contains compounds or DMSO control as shown in steps 3.2–3.4) and mix briefly, either by pipette or on a plate shaker.
(3.8) Cover and incubate the plate at room temperature for 60 minutes and read.
Start time_____
(3.9) Analyze the data: Divide the acceptor/tracer emission (665 nM) by the antibody/donor emission (615 nM) to calculate the TR-FRET emission ratio. Plot the TR-FRET ratio against the log of the test compound concentration and fit to a sigmoidal dose-response curve with a variable slope. Calculate the EC50 concentration from the curve. This is equal to the IC50 value for the inhibitor. Representative data for a set of well-characterized kinase inhibitors are presented in Figure 14.
To prepare 1,500 μL:
(3.10) Calculate the Z’-factor: Using the equation from Zhang et al., calculate the Z’-factor using your negative and positive control wells in column 4.
Z’-factor =  1  –   3 * (σp + σn)
                                   μp – μn
where σp = standard deviation of the positive control wells
σn = standard deviation of the negative control wells
μp = mean of the positive control wells
μn = mean of the negative control wells
(3.11) Assess the data quality: Assays are qualified by the IC50 of the control inhibitor
and by the statistical significance of the assay window as measured by a parameter
known as the Z’-factor. An assay is passing if the IC50 of the control inhibitor is
within ±½ log and is generally considered robust if Z’-factor on the assay window is
greater than 0.5.
Understanding the Z’-factor
Go to...
LT129       1-Jan-2010