48ab - Aplicaciones/Muestras/Productos/Recursos/Póngase en contacto con nosotros
Troubleshooting semiconductor device failure is critical for improving manufacturing yield, reducing costs, and minimizing overall end-of-line test failures. However, isolating faults and defects (e.g. opens, metal shorts, and leakages) in the failure analysis workflow is becoming more challenging due to increasingly complex semiconductor designs.
Optical fault isolation (OFI) is a type of electrical failure analysis that makes use of a variety of optical techniques (photon-emission, static laser stimulation, etc.) to detect the causes of device failure.
This can include both static and dynamic OFI:
Style Sheet for Products Table Specifications
Static OFI techniquesDynamic OFI techniques
Optical beam Induced resistance change (OBIRCH)Laser voltage imaging (LVI)
Optical beam induced current (OBIC)Laser Voltage Probing (LVP)
Light induced voltage alteration (LIVA)Laser voltage tracing (LVT)
Thermally induced voltage alteration (TIVA)Laser-assisted device alteration (LADA)
Static photon emission (PEM)Soft defect localization (SDL)
 Dynamic photon emission (PEM)
Broadly, these techniques allow the user to analyze the performance of electrically active devices and locate critical defects that cause a device to fail.  Understanding these defects, and then eliminating them from the manufacturing process, is crucial if modern fabs are to operate at high yield and profitability.
Thermo Fisher Scientific offers a variety of optical fault isolation systems as part of the Meridian product line. This includes all of the above OFI techniques, encompassing systems for both photon emission and laser stimulation applications. The Meridian product line consists of cost-effective, high-sensitivity solutions for localizing electrical failures in semiconductor devices, and is especially powerful when combined with other analysis workflows and solutions from Thermo Fisher Scientific's portfolio. Click through to the appropriate product pages below for more information.
Semiconductor optical fault isolation workflow example
Resources
Applications
pathfinding_thumb_274x180_144dpi
Desarrollo y trazabilidad de semiconductores
Microscopía electrónica avanzada, haz de iones enfocado y técnicas analíticas asociadas para identificar soluciones viables y métodos de diseño para la fabricación de dispositivos semiconductores de alto rendimiento.
yield_ramp_metrology_2_thumb_274x180
Metrología y rampa de producción
Ofrecemos capacidades analíticas avanzadas para el análisis de defectos, metrología y control de procesos, diseñadas para ayudar a aumentar la productividad y mejorar el rendimiento en una amplia gama de aplicaciones y dispositivos semiconductores.
Análisis de fallos de semiconductores
Análisis de fallos de semiconductores
Las estructuras de dispositivos semiconductores cada vez más complejas dan lugar a que existan más ubicaciones en las que se oculten los defectos inducidos por fallos. Nuestros flujos de trabajo de última generación le ayudarán a localizar y caracterizar los sutiles problemas eléctricos que afectan a la producción, al rendimiento y a la fiabilidad.
physical_characterization_thumb_274x180_144dpi
Caracterización física y química
La demanda continua de los consumidores impulsa la creación de dispositivos electrónicos más pequeños, más rápidos y más baratos. Su producción se basa en instrumentos y flujos de trabajo de alta productividad que generan imágenes, analizan y caracterizan una amplia gama de semiconductores y dispositivos de visualización.
esd_thumb_274x180_144dpi
Cualificación de semiconductor ESD
Se necesita un plan de control de descarga electrostática (ESD) para identificar los dispositivos que son sensibles a ESD. Ofrecemos un conjunto completo de sistemas de prueba para ayudarle con los requisitos de cualificación de su dispositivo.
Análisis de dispositivos semiconductores de energía
Análisis de dispositivos semiconductores de energía
Los dispositivos de energía plantean retos únicos para la localización de fallos, principalmente como resultado de su arquitectura y diseño. Nuestras herramientas y flujos de trabajo de análisis de dispositivos de energía permiten localizar rápidamente los fallos en condiciones de funcionamiento y proporcionar un análisis preciso y de alto rendimiento para la caracterización de materiales, interfaces y estructuras de dispositivos.
Samples
Materiales semiconductores y caracterización de dispositivos
A medida que los dispositivos semiconductores se reducen y se vuelven más complejos, se necesitan nuevos diseños y estructuras. Los flujos de trabajo de análisis en 3D de alta productividad pueden reducir el tiempo de desarrollo de dispositivos, maximizar el rendimiento y garantizar que los dispositivos satisfacen las necesidades futuras del sector.
Más información ›
Style Sheet for Komodo Tabs
Style Sheet to change H2 style to p with em-h2-header class
Products
Style Sheet for Instrument Cards Original
Talos F200E TEM
  • High-quality (S)TEM imaging of semiconductor and microelectronic devices
  • Precise, high-speed chemical characterization with EDS
  • Dedicated semiconductor-related applications
Meridian S System
  • Fault Diagnostic with Active Probe Technology
  • Static laser stimulation (SLS / OBIRCH) and photon emission options
  • Supports both micro-probing and probe card device stimulation
Meridian WS-DP System
  • High-sensitivity, low-noise, low-voltage photon emission detection with broadband DBX or InGaAs camera systems
  • Multi-wavelength laser scanning microscope for scan chain analysis, frequency mapping, transistor probing and isolation of faults
Meridian 7 System
  • Dynamic Optical Fault Isolation for 10nm node and below
  • High resolution visible and Infrared light
  • High-yield sample preparation to 5μm widely available
Meridian IV System
  • High sensitivity extended-wavelength DBX photon emission detection
  • Standard InGaAs photon emission detection
  • Laser Scanning Microscope with multiple wavelength options
Contact us