e14tg2a Cells
Organism Type | Mouse | Mouse ES cells. The feeder-independent ES cell line E14Tg2A grows very quickly and rapidly acidifies the media, turning it yellow in color. When the medium becomes acidified (either by not changing the media every day, or by passaging the cells at too low of a density) the cells may undergo crisis.To prevent cells from differentiating, they should be dissociated down to single cells during their passage. If they are not thoroughly dissociated, they are likely to form large clumps after passage, and the cells within these clumps will then differentiate or die. Similarly, cells should not be plated at very low density, as that may cause them to form large clumps which contain differentiated cells prior to reaching confluence. |
Tissues | Embryo | |
Phenotype | Adherent | |
Primary | no | |
Application | Transient Transfection |
Every Step of the Way, a Wide Range of Cell Health Products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
|
|||