HeLa cells mounted with ProLong® Diamond mountant
HeLa cells were fixed, permeabilized and blocked with BlockAid (B10710), stained with ActinGreen™ 488 ReadyProbes® Reagent (R37110), NucBlue® Live ReadyProbes® Reagent (R37605), and AlexaFluor® 594 Goat Anti-Mouse IgG (H+L) Antibody (A-11005) against ATP Synthase (A-21355), and mounted using ProLong® Diamond mountant (P36961).
HeLa cells were fixed, permeabilized and blocked with BlockAid (B10710), stained with ActinGreen™ 488 ReadyProbes® Reagent (R37110), NucBlue® Live ReadyProbes® Reagent (R37605), and AlexaFluor® 594 Goat Anti-Mouse IgG (H+L) Antibody (A-11005) against ATP Synthase (A-21355), and mounted using ProLong® Diamond mountant (P36961).
Related Products
Related Images