Lysosome Incorporation
(Enlarge)
Fusion of a lysosome with the isolation membrane to form the autolysosome is the penultimate step of the autophagic pathway. Lysosomal membrane markers serve as tools for tracking fusion with the autophagosome prior to degradation of the autolysosome contents, whereas cell permeable LysoTracker® dyes target acidic organelles and are localized within the lysosomes in nanomolar concentrations. Both membrane proteins and Lysotracker dyes serve as effective tools for tracking lysosome behavior.
Autophagy describes the segregation and delivery of cytoplasmic cargo, including proteins and organelles, for degradation by hydrolytic enzymes through the lysosomal machinery. It is critical to the healthy functioning of cells and the failure of autophagy is a major reason for the accumulation of cell damage and aging.
Select Lysosomal Markers
On this page:
Locating and Tracking Lysosomes
Lysosomal membrane markers serve as tools for tracking fusion with the autophagosome prior to degradation of the autolysosome contents. We offer Molecular Probes® CellLight® constructs containing red or green fluorescent proteins for locating and tracking lysosomal membranes.
  • Choice of red or green fluorescent proteins
  • Can be multiplexed with other fluorescent dyes or proteins
See selection guide for Lysosomal Membrane Markers
  	Autophagy Research Human osteosarcoma (U2OS) cells expressing CellLight® Lysosomes-GFP
Human osteosarcoma (U2OS) cells expressing CellLight® Lysosomes-GFP.
Locating and Tracking Lysosomal Contents
Cell-permeable Molecular Probes® LysoTracker® dyes target acidic organelles in nanomolar concentrations. They are available in a range of colors including deep red to facilitate multiplexing with other fluorescent markers.
  • Highly selective for acidic organelles
  • Range of fluorescent colors
  • Can be multiplexed with other fluorescent dyes or proteins
See selection guide for Markers for Lysosomal Contents
  Co-localization using LysoTracker® Deep Red
Co-localization using LysoTracker® Deep Red.
Selection Guides for Lysosomal Markers
Target Lysosomal membranes
Readout Localization of lysosomes by fluorescent protein imaging
Fluorescent label GFP RFP
Standard filter set FITC TRITC
Ex/Em (nm) 485/520 555/584
Signal-to-noise ratio
Photostability
Bibliography Citations
Multiplexing Yes
Live cells Yes
Fixed cells No
Fixable Yes
Format 1 kit 1 kit
Cat. No. C10507 C10504
Target Targets acidic organelles at nanomolar concentrations
Readout Localization of lysosomes by fluorescence imaging
Fluorescent label LysoTracker® Green LysoTracker® Red LysoTracker® Deep Red
Standard filter set FITC TRITC Cy®5
Ex/Em (nm) 504⁄511  577/590 647/668
Signal-to-noise ratio
Photostability
Bibliography Citations
Multiplexing Yes Yes Yes
Live cells Yes Yes Yes
Fixed cells No No No
Fixable No Yes No
Format 20 x 50 uL 20 x 50 uL 5 x 50 uL
Cat. No. L7526 L7528 L12492
Target Lysosomal membranes
Readout Localization of lysosomes by fluorescent protein imaging
Fluorescent label GFP RFP
Standard filter set FITC TRITC
Ex/Em (nm) 485/520 555/584
Signal-to-noise ratio
Photostability
Bibliography Citations
Multiplexing Yes
Live cells Yes
Fixed cells No
Fixable Yes
Format 1 kit 1 kit
Cat. No. C10507 C10504
Target Targets acidic organelles at nanomolar concentrations
Readout Localization of lysosomes by fluorescence imaging
Fluorescent label LysoTracker® Green LysoTracker® Red LysoTracker® Deep Red
Standard filter set FITC TRITC Cy®5
Ex/Em (nm) 504⁄511  577/590 647/668
Signal-to-noise ratio
Photostability
Bibliography Citations
Multiplexing Yes Yes Yes
Live cells Yes Yes Yes
Fixed cells No No No
Fixable No Yes No
Format 20 x 50 uL 20 x 50 uL 5 x 50 uL
Cat. No. L7526 L7528 L12492
Resources